Skip to Content
Merck
All Photos(1)

Key Documents

EMU063061

Sigma-Aldrich

MISSION® esiRNA

targeting mouse C3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACGCCACTATGTCCATCCTGGACATCTCCATGATGACTGGCTTTGCTCCAGACACAAAGGACCTGGAACTGCTGGCCTCTGGAGTAGATAGATACATCTCCAAGTACGAGATGAACAAAGCCTTCTCCAACAAGAACACCCTCATCATCTACCTAGAAAAGATTTCACACACCGAAGAAGACTGCCTGACCTTCAAAGTTCACCAGTACTTTAATGTGGGACTTATCCAGCCCGGGTCGGTCAAGGTCTACTCCTATTACAACCTCGAGGAATCATGCACCCGGTTCTATCATCCAGAGAAGGACGATGGGATGCTCAGCAAGCTGTGCCACAGTGAAATGTGCCGGTGTGCTGAAGAGAACTGCTTCATGCAACAGTCACAGGAGAAGATCAACCTGAATGTCCGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva-Maria Nichols et al.
Kidney international, 88(6), 1314-1322 (2015-07-30)
Abnormal regulation of the complement alternative pathway is associated with C3 glomerulopathy. Complement factor H is the main plasma regulator of the alternative pathway and consists of 20 short consensus repeat (SCR) domains. Although recombinant full-length factor H represents a
Manuela Veglia et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 74(6), 542-552 (2015-09-22)
A threefold higher prevalence of antinuclear antibodies (ANA) has been reported in patients with recurrent pregnancy loss (RPL). Nevertheless, the role of ANA in reproductive failure is still unclear. The aim of this study was to investigate the role of
Hiroyuki Inoshita et al.
PloS one, 8(11), e78736-e78736 (2013-11-14)
The link between glomerular IgA nephropathy (IgAN) and T helper 2 (Th2) response has been implicated, however, the mechanisms are poorly defined because of the lack of an appropriate model. Here we report a novel murine model characterized by lineage-restricted
Masanori A Murayama et al.
Nature communications, 6, 8483-8483 (2015-09-26)
The complement system is important for the host defence against infection as well as for the development of inflammatory diseases. Here we show that C1q/TNF-related protein 6 (CTRP6; gene symbol C1qtnf6) expression is elevated in mouse rheumatoid arthritis (RA) models.
Maria I Fonseca et al.
Journal of neuroinflammation, 8(1), 4-4 (2011-01-18)
Complement proteins and activation products have been found associated with neuropathology in Alzheimer's disease (AD). Recently, a C5a receptor antagonist was shown to suppress neuropathology in two murine models of AD, Tg2576 and 3xTg. Previously, a genetic deficiency of C1q

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service