Skip to Content
Merck
All Photos(1)

Key Documents

EMU000731

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Ywhaz

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCTGCTCTCTTGCAAAAACAGCTTTCGATGAAGCCATTGCTGAACTTGATACATTAAGTGAAGAGTCGTACAAAGACAGCACGCTAATAATGCAGTTACTGAGAGACAACTTAACATTGTGGACATCGGATACCCAAGGAGATGAAGCAGAAGCAGGAGAAGGAGGGGAAAATTAACCGGCCTTCCAACCTTTGTCTGCCTCATTCTAAAATTTACACAGTAGACCATTTGTCATCCATGCTGTCCCACAGATAGTTTTTTTGTTTACGATTTATGACAGGTTTATGTTACTTCTATTTGAATTTCTATATTTCCCATGTGGTTTTTATGTTTTAATATTAGGGGAGTAGAGCCAGTTAACTTTAGGGAGTTACTCATTTTCATCTTGAGGTGGCCAAT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Langyong Mao et al.
American journal of cancer research, 5(6), 1939-1953 (2015-08-14)
The deregulation of microRNAs has been demonstrated in various tumor processes. Here, we report that microRNA-544 (miR-544) is decreased in cervical cancer tissues compared with normal cervical tissues. To identify the mechanisms involved in miR-544 deregulation, we studied the regulation
Gareth E Lim et al.
Nature communications, 6, 7671-7671 (2015-07-30)
The proteins that coordinate complex adipogenic transcriptional networks are poorly understood. 14-3-3ζ is a molecular adaptor protein that regulates insulin signalling and transcription factor networks. Here we report that 14-3-3ζ-knockout mice are strikingly lean from birth with specific reductions in
Hillary E Mulvey et al.
BMC cancer, 15, 571-571 (2015-08-02)
Extracellular vesicles (EVs) are secreted from many cells, carrying cargoes including proteins and nucleic acids. Research has shown that EVs play a role in a variety of biological processes including immunity, bone formation and recently they have been implicated in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service