Skip to Content
Merck
All Photos(1)

Key Documents

EMU000411

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Sirt1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TAAGCGGCTTGAGGGTAATCAATACCTGTTTGTACCACCAAATCGTTACATATTCCACGGTGCTGAGGTATACTCAGACTCTGAAGATGACGTCTTGTCCTCTAGTTCCTGTGGCAGTAACAGTGACAGTGGCACATGCCAGAGTCCAAGTTTAGAAGAACCCTTGGAAGATGAAAGTGAAATTGAAGAATTCTACAATGGCTTGGAAGATGATACGGAGAGGCCCGAATGTGCTGGAGGATCTGGATTTGGAGCTGATGGAGGGGATCAAGAGGTTGTTAATGAAGCTATAGCTACAAGACAGGAATTGACAGATGTAAACTATCCATCAGACAAATCATAACACTATTGAAGCTGTCCGGATTCAGGAATTGCTCCACCAGCATTGGGAACTTTAGCA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hisae Yoshitomi et al.
PloS one, 12(8), e0183988-e0183988 (2017-09-01)
Diabetes is caused by the lack of release or action of insulin. Some foods and supplements can compensate for this deficiency; thus, they can aid in the prevention or treatment of diabetes. The aim of this study was to investigate
Xiwen Xiong et al.
PloS one, 14(2), e0212523-e0212523 (2019-02-23)
Nicotinamide phosphoribosyltransferase (NAMPT) is a rate-limiting enzyme in mammalian nicotinamide adenine dinucleotide (NAD)+ biosynthesis. Through its NAD+-biosynthetic activity, NAMPT influences the activity of NAD+-dependent enzymes, such as sirtuins. NAMPT is able to modulate processes involved in the pathogenesis of non-alcohol
Mickaël Ohanna et al.
Oncotarget, 5(8), 2085-2095 (2014-04-20)
SIRT1 operates as both a tumor suppressor and oncogenic factor depending on the cell context. Whether SIRT1 plays a role in melanoma biology remained poorly elucidated. Here, we demonstrate that SIRT1 is a critical regulator of melanoma cell proliferation. SIRT1
Jie-Ning Zhu et al.
Scientific reports, 7(1), 11879-11879 (2017-09-21)
The molecular mechanisms underlying anthracyclines-induced cardiotoxicity have not been well elucidated. MiRNAs were revealed dysregulated in the myocardium and plasma of rats received Dox treatment. MicroRNA-34a-5p (miR-34a-5p) was verified increased in the myocardium and plasma of Dox-treated rats, but was
Yuanbang Lin et al.
Journal of natural medicines, 72(3), 715-723 (2018-04-24)
Berberine (BBR) has been demonstrated to protect against renal ischemia/reperfusion injury; however, the underlying molecular mechanism is largely unknown. In the present study, we examined the role of silent information regulator 1 (Sirt1)/p53 in the protective effect of BBR on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service