Skip to Content
Merck

Skip To

EMNC013301

MISSION® esiRNA

targeting mouse Fign

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
$287.00
50 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
$517.00

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324

$287.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTCGCCCATCTCTGAGAAACATGTTAGCATTCCCAAGTATTTTACACTGGCAATCAGACTACTTAAGACAAACAAATAGAAACATGTGTTACATTCTTTTGACTGACACCCTCACTTCCTAAGTTTAAGAGAAACATAGTACTTAATGATTGCTGACTGCAGTCTTTGGTGGTTTTAAATGCCTTCCTTTTAGTTCTGTGACTCCTGATATCAGTGAATGGAAACACCACCATTGCACAGGCCTGCACAACACCAAAATAAAATAAACCAAAACCAAAACCAAAACAAAACAAAAAAACAAAATTAAAAATGGCACTTCCCAAAGGAATTAACACACATGGAGTCATGGTGCTTCGAAGTTCCCTATATGAAAACATTTTTTTTTGTTTCCTATTTCCCTTTGTAACATGTATTTGTTCCTTAAAGTACTGTACAACCAATGGCACATTGAA

GenBank accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

mouse ... FIGN(60344)

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EMNC000671EMNC000071EMNC000661
Gene Information

mouse ... FIGN(60344)

Gene Information

mouse ... 4833419E13Rik(74579)

Gene Information

mouse ... AI450353(103729)

Gene Information

mouse ... 2610030P05Rik(69258)

esiRNA cDNA target sequence

CTCGCCCATCTCTGAGAAACATGTTAGCATTCCCAAGTATTTTACACTGGCAATCAGACTACTTAAGACAAACAAATAGAAACATGTGTTACATTCTTTTGACTGACACCCTCACTTCCTAAGTTTAAGAGAAACATAGTACTTAATGATTGCTGACTGCAGTCTTTGGTGGTTTTAAATGCCTTCCTTTTAGTTCTGTGACTCCTGATATCAGTGAATGGAAACACCACCATTGCACAGGCCTGCACAACACCAAAATAAAATAAACCAAAACCAAAACCAAAACAAAACAAAAAAACAAAATTAAAAATGGCACTTCCCAAAGGAATTAACACACATGGAGTCATGGTGCTTCGAAGTTCCCTATATGAAAACATTTTTTTTTGTTTCCTATTTCCCTTTGTAACATGTATTTGTTCCTTAAAGTACTGTACAACCAATGGCACATTGAA

esiRNA cDNA target sequence

CATGGGCACTGGTTCTAGGTCAGATCAGTGACAAGTTCAAGGCTCTAGCACTTAGTCACTATTGAAACCACGGGTCAAATACAGTTCACAGGGAAGGAAGACTGTCTTCTGTTGGAGGAGGGAGACATGTCTACACTAGTGATCATACTAGTGAATGTTGTTCTAGATAGCAGACCCAGGGAGGGGATGCAGAGGACACAGGTCTGGAGAAATAGCTCAGCAGAAGAACCCTGACTACTCTTCCAGAGGCCCC

esiRNA cDNA target sequence

GACCTGGTATTGCCATTGCTTTGGCGCTGGCTCTGACCAGGGTGTGACCCCTTAATCTTCTCTCTGAGCTGATTCTGCTGACCACATGGGGGTATTTGAACCTATGTGGATCCTGGCTTATAATGTGTAAGAGTATTTCTTTTGTATCCTTTGAATTACTGATACTTTCATGTTTTCCTTCTTAGGTCCAGGTGAGAAATCAGGGTTGAAAACTGCATG

esiRNA cDNA target sequence

CAACCTTGCAAGACATTGGACAGTGTTTTCTGAGAAGTACTCAATGAATATCTGAATTAGTAGATGTCTTAGTTAGGGTTCTATTGCTGTGAAGAGACACCATGACCAAGGCAACTCTTACAAAGGAAAACATTTAATTAGGGCTGGCTTACAGTTGCAGAGGCTTAGCTCATTTTTGTCATGGCAGGAAGCATGGCAGCATGCAGGCAGACATAGGGCTGGACAAGGAGCT

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

NCBI accession no.

NM_021716.4

NCBI accession no.

-

NCBI accession no.

NR_028364.1

NCBI accession no.

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters