Skip to Content
Merck
All Photos(1)

Key Documents

EHU218961

Sigma-Aldrich

MISSION® esiRNA

targeting human CEBPA

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$534.00
20 μG
$534.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
50 μG
$534.00
20 μG
$534.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCGAGTCACACCAGAAAGCTAGGTCGTGGGTCAGCTCTGAGGATGTATACCCCTGGTGGGAGAGGGAGACCTAGAGATCTGGCTGTGGGGCGGGCATGGGGGGTGAAGGGCCACTGGGACCCTCAGCCTTGTTTGTACTGTATGCCTTCAGCATTGCCTAGGAACACGAAGCACGATCAGTCCATCCCAGAGGGACCGGAGTTATGACAAGCTTTCCAAATATTTTGCTTTATCAGCCGATATCAACACTTGTATCTGGCCTCTGTGCCCCAGCAGTGCCTTGTGCAATGTGAATGTGCGCGTCTCTGCTAAACCACCATTTTATTTGGTTTTTGTTTTGTTTTGGTTTTGCTCGGATACTTGCCAAAATGAGACTCTCCGTCGGCAGCTGGGGGAAGGGTCTGAGACTCCCTTTCCTTTTGGTTTTGGGATTACTTTTGATCCTGGGGGACCAATGAGGTGAGGGGGGTTCTCCTTTGCCCTCAGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Rita Lourenço et al.
Nature communications, 11(1), 785-785 (2020-02-09)
Extracellular signals such as TGF-β can induce epithelial-to-mesenchymal transition (EMT) in cancers of epithelial origin, promoting molecular and phenotypical changes resulting in pro-metastatic characteristics. We identified C/EBPα as one of the most TGF-β-mediated downregulated transcription factors in human mammary epithelial
Wei Liu et al.
Respiratory research, 20(1), 281-281 (2019-12-13)
Fibroblasts regulate tissue homeostasis and the balance between tissue repair and fibrosis. CCAAT/enhancer-binding protein alpha (CEBPA) is a key transcription factor that regulates adipogenesis. CEBPA has been shown to be essential for lung maturation, and deficiency of CEBPA expression leads
Rui Su et al.
Cell, 172(1-2), 90-105 (2017-12-19)
R-2-hydroxyglutarate (R-2HG), produced at high levels by mutant isocitrate dehydrogenase 1/2 (IDH1/2) enzymes, was reported as an oncometabolite. We show here that R-2HG also exerts a broad anti-leukemic activity in vitro and in vivo by inhibiting leukemia cell proliferation/viability and by promoting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service