Skip to Content
Merck
All Photos(1)

Key Documents

EHU154611

Sigma-Aldrich

MISSION® esiRNA

targeting human KCNA3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGTTCTCCTTCGAACTGCTGGTGCGGTTCTTCGCTTGTCCTAGCAAAGCCACCTTCTCGCGAAACATCATGAACCTGATCGACATTGTGGCCATCATTCCTTATTTTATCACTCTGGGTACCGAGCTGGCCGAACGACAGGGCAATGGACAGCAGGCCATGTCTCTGGCCATCCTGAGGGTCATCCGCCTGGTAAGGGTCTTCCGCATCTTCAAGCTGTCGCGCCACTCCAAGGGGCTGCAGATCCTCGGGCAAACGCTGAAGGCGTCCATGCGGGAGCTGGGATTGCTCATCTTCTTCCTCTTTATTGGGGTCATCCTTTTCTCCAGCGCGGTCTACTTTGCCGAGGCAGACGACCCCACTTCAGGTTTCAGCAGCATCCCGGATGCCTTCTGGTGGGCAGTGGTAACCATGACAACAGTGGGTTACGGCGATATGCACCCAGTGACCATAGGGGGCAAGATTGTGGGATCTCTCTGTGCCATCGCCGGTGTCTTGACCATCGCATTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Hong Kan et al.
Archives of biochemistry and biophysics, 591, 150-156 (2016-01-10)
Ion channels expressed in macrophages have been tightly related to atherosclerosis by coupling cellular function. How the voltage-gated potassium channels (Kv) affect macrophage migration remain unknown. The aim of our study is to investigate whether Kv1.3-ERK signaling pathway plays an
Marat Khodoun et al.
Science advances, 6(47) (2020-11-20)
Lupus nephritis (LN) is an autoimmune disease with substantial morbidity/mortality and limited efficacy of available therapies. Memory T (Tm) lymphocytes infiltrate LN kidneys, contributing to organ damage. Analysis of LN, diabetic nephropathy, and healthy donor kidney biopsies revealed high infiltration
Roberta Peruzzo et al.
Redox biology, 37, 101705-101705 (2020-10-03)
The potassium channel Kv1.3, involved in several important pathologies, is the target of a family of psoralen-based drugs whose mechanism of action is not fully understood. Here we provide evidence for a physical interaction of the mitochondria-located Kv1.3 (mtKv1.3) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service