Skip to Content
Merck
All Photos(1)

Key Documents

EHU153321

Sigma-Aldrich

MISSION® esiRNA

targeting human CCND1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AACTACCTGGACCGCTTCCTGTCGCTGGAGCCCGTGAAAAAGAGCCGCCTGCAGCTGCTGGGGGCCACTTGCATGTTCGTGGCCTCTAAGATGAAGGAGACCATCCCCCTGACGGCCGAGAAGCTGTGCATCTACACCGACAACTCCATCCGGCCCGAGGAGCTGCTGCAAATGGAGCTGCTCCTGGTGAACAAGCTCAAGTGGAACCTGGCCGCAATGACCCCGCACGATTTCATTGAACACTTCCTCTCCAAAATGCCAGAGGCGGAGGAGAACAAACAGATCATCCGCAAACACGCGCAGACCTTCGTTGCCCTCTGTGCCACAGATGTGAAGTTCATTTCCAATCCGCCCTCCATGGTGGCAGCGGGGAGCGTGGTGGCCGCAGTGCAAGGCCTGAACCTGAGGAGCCCCAACAACTTCCTGTCCTACTACCGCCTCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohong Jiang et al.
Gene therapy, 27(12), 557-566 (2020-06-07)
LncRNAs are reported to participate in the progression of various diseases including diabetic nephropathy. Currently, we reported that SNHG16 was obviously upregulated in db/db mice and high glucose-treated mice mesangial cells. Then, functional experiments showed that SNHG16 silencing significantly inhibited
Chuanyong Wu et al.
Journal of Cancer, 11(7), 1959-1967 (2020-03-21)
Accumulating evidences showed that aberrantly expressed long noncoding RNAs (lncRNAs) have critical roles in many cancers. However, the expression and roles of a poorly studied lncRNA PCNA-AS1 in non-small-cell lung cancer (NSCLC) remain unknown. In this study, we investigated the
Mingming Wang et al.
Journal of cellular physiology, 235(2), 1588-1600 (2019-07-17)
Prostate cancer (PCa) is one of the major health problems of the aging male. The roles of dysregulated microRNAs in PCa remain unclear. In this study, we mined the public published data and found that miR-487a-3p was significantly downregulated in
Dexin Yin et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 49(4), 1499-1511 (2018-09-12)
Recent studies have suggested that several lncRNAs contribute to the angiogenic function of endothelial cells. Herein, we set out to reveal whether lncRNA UCA1 has functions in endothelial angiogenesis. The expression levels of lncRNA UCA1, miR-195 and CCND1 in human
Hao Ding et al.
Journal of pharmacological sciences, 143(3), 219-225 (2020-05-19)
Chemoresistance is one of the major barriers in chemotherapy-based hepatocellular carcinoma (HCC) intervention. 5-Fluorouracil (5-Fu) is a widely used as an anticancer drug. Liver cancer stem cells (LCSCs) are considered the origin of tumor recurrence and resistance. CCND1 (Cyclin D1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service