Skip to Content
Merck
All Photos(1)

Key Documents

EHU151371

Sigma-Aldrich

MISSION® esiRNA

targeting human MDK

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCTGCAACTGGAAGAAGGAGTTTGGAGCCGACTGCAAGTACAAGTTTGAGAACTGGGGTGCGTGTGATGGGGGCACAGGCACCAAAGTCCGCCAAGGCACCCTGAAGAAGGCGCGCTACAATGCTCAGTGCCAGGAGACCATCCGCGTCACCAAGCCCTGCACCCCCAAGACCAAAGCAAAGGCCAAAGCCAAGAAAGGGAAGGGAAAGGACTAGACGCCAAGCCTGGATGCCAAGGAGCCCCTGGTGTCACATGGGGCCTGGCCCACGCCCTCCCTCTCCCAGGCCCGAGATGTGACCCACCAGTGCCTTCTGTCTGCTCGTTAGCTTTAATCAATCATGCCCTGCCTTGTCCCTCTCACTCCCCAGCCCCACCCCTAAGTGCCCAAAGTGGGGAGGGACAAGGGATTCTGGGAAGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Lu et al.
Journal of experimental & clinical cancer research : CR, 37(1), 147-147 (2018-07-14)
Exosomes are small vesicles containing a wide range of functional proteins, mRNA and miRNA. Exosomal miRNAs from cancer cells play crucial roles in mediating cell-cell communication and tumor-microenvironment cross talk, specifically in enabling metastasis and promoting angiogenesis. We focused on
Bin Sun et al.
Oncotarget, 8(20), 32523-32535 (2017-04-22)
Midkine is overexpressed in hepatocellular carcinoma (HCC) and plays a role in tumor progression, but less is known about its role in resistance of circulating tumor cells (CTCs) to anoikis which leading to recurrence and metastasis. The aim of the
Huilian Huang et al.
Clinical laboratory, 61(10), 1501-1508 (2015-12-09)
Mounting evidence indicates that nuclear targeting by growth factors plays an indispensable role on their biological activities. Midkine (MK) is a multifunctional growth factor and has been discovered to play important roles in carcinogenesis. MK has been reported to localize

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service