Skip to Content
Merck
All Photos(1)

Key Documents

EHU151281

Sigma-Aldrich

MISSION® esiRNA

targeting human PCSK6 (2)

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCCCCAAATTATGATTCCTACGCCAGCTACGACGTGAACGGCAATGATTATGACCCATCTCCACGATATGATGCCAGCAATGAAAATAAACACGGCACTCGTTGTGCGGGAGAAGTTGCTGCTTCAGCAAACAATTCCTACTGCATCGTGGGCATAGCGTACAATGCCAAAATAGGAGGCATCCGCATGCTGGACGGCGATGTCACAGATGTGGTCGAGGCAAAGTCGCTGGGCATCAGACCCAACTACATCGACATTTACAGTGCCAGCTGGGGGCCGGACGACGACGGCAAGACGGTGGACGGGCCCGGCCGACTGGCTAAGCAGGCTTTCGAGTATGGCATTAAAAAGGGCCGGCAGGGCCTGGGCTCCATTTTCGTCTGGGCATCTGGGAATGGCGGGAGAGAGGGGGACTACTGCTCGTGCGATGGCTACACCAACAGCATCTACACCATCTCCGTCAGCAGCGCCACCGAGAATGGCTACAAGCCCTGGTACCTGGAAGAGTGTGCCTCCACCCTGGCCACCACCTACAGCAGTGGGGCCTTTTATGAGCGAAAAATCGTCACC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Feng Tian et al.
Molecular medicine reports, 14(6), 5205-5210 (2016-10-26)
Previous studies have demonstrated the overexpression of paired basic amino acid cleaving enzyme 4 (PACE4) mRNA in prostate cancer tissues. This overexpression is correlated with higher circulating protein levels in certain patients, however, the role of PACE4 in apoptosis and the
Jian Fu et al.
Molecular carcinogenesis, 54(9), 698-706 (2014-01-18)
Proprotein convertases (PC), a family of serine proteases, process cancer-related substrates such as growth factors, growth factor receptors, cell adhesion molecules, metalloproteinases, etc. HIF-1α is a major transcription factor involved in tumorigenesis by sensing intratumoral hypoxia. Furin (PCSK3) is one
Zhiyong Yao et al.
Drug design, development and therapy, 9, 5911-5923 (2015-11-26)
PACE4 is a proprotein convertase capable of processing numerous substrates involved in tumor growth, invasion, and metastasis. However, the precise role of PACE4 during prostate cancer cell apoptosis has not been reported. In the present study, human prostate cancer cell
Huiyu Jiang et al.
Molecular medicine reports, 12(5), 7681-7686 (2015-10-16)
The aim of the present study was to assess the effects of pro-protein convertase subtilisin/kexin type 6 (PCSK6), a proteinase implicated in the proteolytic activity of various precursor proteins and involved in the regulation of protein maturation, in fibroblast‑like synoviocytes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service