Skip to Content
Merck
All Photos(1)

Documents

EHU149011

Sigma-Aldrich

MISSION® esiRNA

targeting human SLC5A2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTGCTGCATATTTCCTGCTGGTCATTGGCGTTGGCTTGTGGTCCATGTGCAGAACCAACAGAGGCACTGTGGGCGGCTACTTCCTGGCAGGACGCAGCATGGTGTGGTGGCCGGTTGGGGCCTCTCTCTTCGCCAGCAACATCGGCAGTGGCCACTTTGTGGGCCTGGCAGGGACTGGCGCTGCAAGTGGCTTGGCTGTTGCTGGATTCGAGTGGAATGCGCTCTTCGTGGTGCTGCTACTGGGCTGGCTGTTTGCACCCGTGTACCTGACAGCGGGGGTCATCACGATGCCACAGTACCTGCGCAAGCGCTTCGGCGGCCGCCGCATCCGCCTCTACCTGTCTGTGCTCTCCCTTTTCCTGTACATCTTCACCAAGATCTCAGTGGACATGTTCTCCGGAGCTGTATTCATCCAGCAGGCTCTGGGCTGGAACATCTA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Ida-Yonemochi et al.
Journal of dental research, 99(8), 977-986 (2020-04-30)
Glucose is an essential source of energy for mammalian cells and is transported into the cells by glucose transporters. There are 2 types of glucose transporters: one is a passive glucose transporter, GLUT (SLC2A), and the other is a sodium-dependent
Jinpeng Li et al.
JCI insight, 5(6) (2020-03-07)
Sodium glucose cotransporter 2 (SGLT2) inhibitors are beneficial in halting diabetic kidney disease; however, the complete mechanisms have not yet been elucidated. The epithelial-mesenchymal transition (EMT) is associated with the suppression of sirtuin 3 (Sirt3) and aberrant glycolysis. Here, we
Jin Hee Kim et al.
Diabetes, obesity & metabolism, 22(3), 373-382 (2019-11-07)
To investigate the effect of dapagliflozin, a sodium-glucose co-transporter-2 (SGLT2) inhibitor, on renal gluconeogenesis in vitro, ex vivo and in vivo. We treated HK-2 cells (human renal proximal tubule cells) and mouse primary renal proximal tubule cells with dapagliflozin, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service