Skip to Content
Merck
All Photos(1)

Documents

EHU144201

Sigma-Aldrich

MISSION® esiRNA

targeting human SLC2A2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCAGCTACCGACAGCCTATTCTAGTGGCACTGATGCTGCATGTGGCTCAGCAATTTTCCGGAATCAATGGCATTTTTTACTACTCAACCAGCATTTTTCAGACGGCTGGTATCAGCAAACCTGTTTATGCAACCATTGGAGTTGGCGCTGTAAACATGGTTTTCACTGCTGTCTCTGTATTCCTTGTGGAGAAGGCAGGGCGACGTTCTCTCTTTCTAATTGGAATGAGTGGGATGTTTGTTTGTGCCATCTTCATGTCAGTGGGACTTGTGCTGCTGAATAAGTTCTCTTGGATGAGTTATGTGAGCATGATAGCCATCTTCCTCTTTGTCAGCTTCTTTGAAATTGGGCCAGGCCCGATCCCCTGGTTCATGGTGGCTGAGTTTTTCAGTCAAGGACCACGTCCTGCTGCTTTAGCAATAGCTGCATTCAGCAATTGGACCTGCAATTTCATTGTAGCTCTGTGTTTCCAGTACATTGCGGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Yong Lee et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 35(1), e21236-e21236 (2020-12-19)
Cadmium (Cd) is an environmental contaminant that causes renal toxicity. We have previously demonstrated that Cd induces renal toxicity by altering transcriptional activities. In this study, we show that Cd markedly inhibited the activity of transcription factor MEF2A in HK-2
Qiao Zhang et al.
Diabetes research and clinical practice, 142, 363-373 (2018-06-26)
High levels of circulating free fatty acids (FFAs), inflammation and oxidative stress are important causes for insulin resistance (IR) and type 2 diabetes mellitus. The aim of this study was to investigate the mechanisms of EGCG in alleviating IR in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service