Skip to Content
Merck
All Photos(1)

Key Documents

EHU131401

Sigma-Aldrich

MISSION® esiRNA

targeting human IGFBP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGCCAAACTGCAACAAGAATGGATTTTATCACAGCAGACAGTGTGAGACATCCATGGATGGAGAGGCGGGACTCTGCTGGTGCGTCTACCCTTGGAATGGGAAGAGGATCCCTGGGTCTCCAGAGATCAGGGGAGACCCCAACTGCCAGATATATTTTAATGTACAAAACTGAAACCAGATGAAATAATGTTCTGTCACGTGAAATATTTAAGTATATAGTATATTTATACTCTAGAACATGCACATTTATATATATATGTATATGTATATATATATAGTAACTACTTTTTATACTCCATACATAACTTGATATAGAAAGCTGTTTATTTATTCACTGTAAGTTTATTTTTTCTACACAGTAAAAACTTGTACTATGTTAATAACTTGTCCTATGTCAATTTGTATATCATGAAACACTTCTCATCATATTGTATGTAAGTAATTGCATTTCTGCTCTTCCAAAGCTCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Zheng et al.
Experimental & molecular medicine, 50(9), 121-121 (2018-09-14)
β-Elemene, an active component of natural plants, has been shown to exhibit anticancer properties. However, the detailed mechanism underlying these effects has yet to be determined. In this study, we show that β-elemene inhibits the growth of lung cancer cells.
Hideno Tochigi et al.
Scientific reports, 7, 40001-40001 (2017-01-05)
Endometrial decidualization represents an essential step for the successful implantation of the embryo; however, the molecular mechanism behind this differentiation process remains unclear. This study aimed to identify novel microRNAs (miRNAs) involved in the regulation of decidual gene expression in
Ali Vaziri-Gohar et al.
Molecular and cellular endocrinology, 422, 160-171 (2015-12-23)
Tamoxifen, a selective estrogen receptor modulator, is a commonly prescribed adjuvant therapy for estrogen receptor-α (ERα)-positive breast cancer patients. To determine if extracellular factors contribute to the modulation of IGF-1 signaling after tamoxifen treatment, MCF-7 cells were treated with IGF-1 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service