Skip to Content
Merck
All Photos(1)

Key Documents

EHU130241

Sigma-Aldrich

MISSION® esiRNA

targeting human FOXP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAAATGGTGTCTGCAAGTGGCCCGGATGTGAGAAGGTCTTCGAAGAGCCAGAGGACTTCCTCAAGCACTGCCAGGCGGACCATCTTCTGGATGAGAAGGGCAGGGCACAATGTCTCCTCCAGAGAGAGATGGTACAGTCTCTGGAGCAGCAGCTGGTGCTGGAGAAGGAGAAGCTGAGTGCCATGCAGGCCCACCTGGCTGGGAAAATGGCACTGACCAAGGCTTCATCTGTGGCATCATCCGACAAGGGCTCCTGCTGCATCGTAGCTGCTGGCAGCCAAGGCCCTGTCGTCCCAGCCTGGTCTGGCCCCCGGGAGGCCCCTGACAGCCTGTTTGCTGTCCGGAGGCACCTGTGGGGTAGCCATGGAAACAGCACATTCCCAGAGTTCCTCCACAACATGGACTACTTCAAGTTCCACAACATGCGACCCCCTTTCACCTACGCCACGCTCATCCGCTGGGCCATCCTGGAGGCTCCAGAGAAGCAGCGGACACTCAAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grethe Skretting et al.
Journal of cellular biochemistry, 120(8), 12924-12936 (2019-03-13)
Single nucleotide polymorphisms (SNPs) may play an important role in the risk of certain diseases. We have previously shown that the -287T/C SNP of the tissue factor pathway inhibitor (TFPI) gene promoter region exerts differential impact on TFPI mRNA expression;
Dong Fan et al.
Oncology letters, 20(6), 292-292 (2020-10-27)
Forkhead box P3 (FOXP3), an X-linked tumor suppressor gene, plays an important role in breast cancer. However, the biological functions of FOXP3 in breast cancer apoptosis remain unclear. To investigate the underlying genes and networks regulated by FOXP3 in breast
Hanwei Zhang et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 22(21), 5349-5361 (2016-11-03)
The transcriptional regulation mediating cancer cell differentiation into distinct molecular subtypes and modulating sensitivity to existing treatments is an enticing therapeutic target. Our objective was to characterize the ability of the forkhead/winged transcription factor FOXP3 to modulate the differentiation of
Chun Li et al.
Pathology, research and practice, 213(10), 1251-1256 (2017-09-25)
Our study aimed to investigate the biological role of FOXP3 expression in human lung adenocarcinoma (LAD) tissues and evaluate its involvement in cell proliferation and chemosensitivity to cisplatin in LAD cells. Paraffin-embedded tissues from 50 LAD patients were collected to
Maria Napolitano et al.
Oncotarget, 6(10), 8261-8270 (2015-04-01)
Short-course preoperative radiotherapy (SC-RT) followed by total mesorectal excision (TME) is one therapeutic option for locally advanced rectal cancer (LARC) patients. Since radio-induced DNA damage may affect tumor immunogenicity, Myeloid-derived suppressor cells (MDSCs) and T regulatory cells (Tregs) were evaluated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service