Skip to Content
Merck
All Photos(1)

Key Documents

EHU130211

Sigma-Aldrich

MISSION® esiRNA

targeting human PRDM1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCAGCTCTCCAATCTGAAGGTCCACCTGAGAGTGCACAGTGGAGAACGGCCTTTCAAATGTCAGACTTGCAACAAGGGCTTTACTCAGCTCGCCCACCTGCAGAAACACTACCTGGTACACACGGGAGAAAAGCCACATGAATGCCAGGTCTGCCACAAGAGATTTAGCAGCACCAGCAATCTCAAGACCCACCTGCGACTCCATTCTGGAGAGAAACCATACCAATGCAAGGTGTGCCCTGCCAAGTTCACCCAGTTTGTGCACCTGAAACTGCACAAGCGTCTGCACACCCGGGAGCGGCCCCACAAGTGCTCCCAGTGCCACAAGAACTACATCCATCTCTGTAGCCTCAAGGTTCACCTGAAAGGGAACTGCGCTGCGGCCCCGGCGCCTGGGCTGCCCTTGGAAGATCTGACCCGAATC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liuluan Zhu et al.
Journal of hematology & oncology, 10(1), 124-124 (2017-06-21)
T cell immunoglobulin and immunoreceptor tyrosine-based inhibitory motif (ITIM) domain (TIGIT) and programmed cell death protein 1 (PD-1) are important inhibitory receptors that associate with T cell exhaustion in acute myeloid leukemia (AML). In this study, we aimed to determine
Gundappa Saha et al.
Frontiers in cellular and infection microbiology, 10, 594431-594431 (2020-11-17)
Precise regulation of inflammasome is critical during any pathogenic encounter. The whole innate immune system comprising of pattern recognition receptors (PRRs) relies on its ability to sense microbes. The fate of cellular death in infected cells depends mostly on the
Enzo Acerbi et al.
Scientific reports, 6, 23128-23128 (2016-03-16)
T helper 17 (TH17) cells represent a pivotal adaptive cell subset involved in multiple immune disorders in mammalian species. Deciphering the molecular interactions regulating TH17 cell differentiation is particularly critical for novel drug target discovery designed to control maladaptive inflammatory
Woo-Shin Kim et al.
Scientific reports, 7(1), 10626-10626 (2017-09-08)
Transglutaminase 2 (TG2) performs multiple reactions, including transamidation, and also plays a role in signal transduction as a GTP-binding protein. In this study, we reveal that TG2 controls osteoclast differentiation and bone homeostasis in mice. Osteoclasts specifically expressed the TG2
Prontip Saelee et al.
Frontiers in immunology, 8, 383-383 (2017-04-26)
The transcription factor Ets1 is highly expressed in B lymphocytes. Loss of Ets1 leads to premature B cell differentiation into antibody-secreting cells (ASCs), secretion of autoantibodies, and development of autoimmune disease. Despite the importance of Ets1 in B cell biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service