Skip to Content
Merck
All Photos(1)

Key Documents

EHU125061

Sigma-Aldrich

MISSION® esiRNA

targeting human CALCA

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCATGTGGTTTGGTTCCTCTCTGGTGGCTCTTTGGGCTGGTATTGGTGGCTTTCCTTGTGGCAGAGGATGTCTCAAACTTCAGATGGGAGGAAAGAGAGCAGGACTCACAGGTTGGAAGAGAATCACCTGGGAAAATACCAGAAAATGAGGGCCGCTTTGAGTCCCCCAGAGATGTCATCAGAGCTCCTCTGTCCTGCTTCTGAATGTGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanjun Guo et al.
Peptides, 121, 170121-170121 (2019-08-07)
Endothelial dysfunction is considered to be an initial indicator in diabetes-induced macrovascular complications. Evidence has shown that CGRP is an important neuropeptide active in vascular system, especially in vasorelaxation. This study aimed to investigate the role of CGRP in high-glucose-induced
Yin-Hung Chu et al.
Cells, 9(3) (2020-03-19)
Epithelial-mesenchymal transition (EMT) is strongly correlated with tumor metastasis and contains several protein markers, such as E-cadherin. Carbonic anhydrase III (CA III) exhibits low carbon dioxide hydratase activity in cancer. However, the detailed mechanisms of CA III and their roles
Richard A Byrd et al.
Endocrinology, 156(7), 2417-2428 (2015-04-11)
The tumorigenic potential of dulaglutide was evaluated in rats and transgenic mice. Rats were injected sc twice weekly for 93 weeks with dulaglutide 0, 0.05, 0.5, 1.5, or 5 mg/kg corresponding to 0, 0.5, 7, 20, and 58 times, respectively
Ines Block et al.
Oncogene, 38(23), 4560-4573 (2019-02-14)
Breast cancer is a heterogeneous genetic disease driven by the accumulation of individual mutations per tumor. Whole-genome sequencing approaches have identified numerous genes with recurrent mutations in primary tumors. Although mutations in well characterized tumor suppressors and oncogenes are overrepresented
Annachiara Sarnella et al.
International journal of molecular sciences, 21(21) (2020-11-14)
Cell plasticity is the ability that cells have to modify their phenotype, adapting to the environment. Cancer progression is under the strict control of the the tumor microenvironment that strongly determines its success by regulating the behavioral changes of tumor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service