Skip to Content
Merck
All Photos(1)

Key Documents

EHU119241

Sigma-Aldrich

MISSION® esiRNA

targeting human CAMK4

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTTCTTCGCCTCTCACATCCAAACATTATAAAACTTAAAGAGATATTTGAAACCCCTACAGAAATCAGTCTGGTCCTAGAACTCGTCACAGGAGGAGAACTGTTTGATAGGATTGTGGAAAAGGGATATTACAGTGAGCGAGATGCTGCAGATGCCGTTAAACAAATCCTGGAGGCAGTTGCTTATCTACATGAAAATGGGATTGTCCATCGTGATCTCAAACCAGAGAATCTTCTTTATGCAACTCCAGCCCCAGATGCACCACTCAAAATCGCTGATTTTGGACTCTCTAAAATTGTGGAACATCAAGTGCTCATGAAGACAGTATGTGGAACCCCAGGGTACTGCGCACCTGAAATTCTTAGAGGTTGTGCCTATGGACCTGAGGTGGACATGTGGTCTGTAGGAATAATCACCTACATCTTACTTTGTGGATTTGAACCATTCTATGATGAAAGAGGCGATCAGTTCATGTTCAGGAGAATTCTGAATTGTGAATATTACTTTATCTCCCCCTGGTGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DanDan Shi et al.
Inflammation, 41(1), 199-212 (2017-10-04)
The objective of this study is to investigate the role of Calmodulin-dependent protein kinase IV (CaMKIV) in Cardiotoxin (CTX)-induced mice muscle inflammation. CTX injection i.m. was performed to induce B6 mice acute tibialis anterior (TA) muscle injury. The mice were
Kunihiro Ichinose et al.
Arthritis & rheumatology (Hoboken, N.J.), 68(4), 944-952 (2015-12-05)
Kidney podocytes and their slit diaphragms prevent urinary protein loss. T cells from patients with systemic lupus erythematosus display increased expression of calcium/calmodulin-dependent protein kinase IV (CaMKIV). The present study was undertaken to investigate the role of CaMKIV in podocyte
Kayaho Maeda et al.
The Journal of clinical investigation, 128(8), 3445-3459 (2018-07-10)
Podocyte malfunction occurs in autoimmune and nonautoimmune kidney disease. Calcium signaling is essential for podocyte injury, but the role of Ca2+/calmodulin-dependent kinase (CaMK) signaling in podocytes has not been fully explored. We report that podocytes from patients with lupus nephritis
Xueyuan Zhou et al.
Immunology, 143(2), 287-299 (2014-04-30)
Prostaglandin E2 (PGE2 ) is an important inducer of inflammation, which is also closely linked to the progress of tumours. In macrophages, PGE2 production is regulated by arachidonic acid release and cyclooxygenase-2 (COX-2) expression. In the present study, we found

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service