Skip to Content
Merck
All Photos(1)

Key Documents

EHU114701

Sigma-Aldrich

MISSION® esiRNA

targeting human CD46

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGTCTCAGATGACGCCTGTTATAGAGAAACATGTCCATATATACGGGATCCTTTAAATGGCCAAGCAGTCCCTGCAAATGGGACTTACGAGTTTGGTTATCAGATGCACTTTATTTGTAATGAGGGTTATTACTTAATTGGTGAAGAAATTCTATATTGTGAACTTAAAGGATCAGTAGCAATTTGGAGCGGTAAGCCCCCAATATGTGAAAAGGTTTTGTGTACACCACCTCCAAAAATAAAAAATGGAAAACACACCTTTAGTGAAGTAGAAGTATTTGAGTATCTTGATGCAGTAACTTATAGTTGTGATCCTGCACCTGGACCAGATCCATTTTCACTTATTGGAGAGAGCACGATTTATTGTGGTGACAATTCAGTGTGGAGTCGTGCTGCTCCAGAGTGTA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn R Stein et al.
Nature communications, 10(1), 2699-2699 (2019-06-22)
Human cytomegalovirus (CMV) causes a wide array of disease to diverse populations of immune-compromised individuals. Thus, a more comprehensive understanding of how CMV enters numerous host cell types is necessary to further delineate the complex nature of CMV pathogenesis and
Pei Qiao et al.
Scientific reports, 7(1), 145-145 (2017-03-10)
Bullous pemphigoid (BP) is an autoimmune bullous disease caused by autoantibodies against BP180 in the epidermal basement membrane. Autoantibody-mediated complement activation is an important process in BP pathogenesis. CD46, a crucial complement regulatory protein in the complement activation, has been
Meng-Lay Lin et al.
Oncotarget, 6(25), 21685-21703 (2015-08-19)
The Nuclear Receptor (NR) superfamily of transcription factors comprises 48 members, several of which have been implicated in breast cancer. Most important is estrogen receptor-α (ERα), which is a key therapeutic target. ERα action is facilitated by co-operativity with other
J Song et al.
Cell death & disease, 6, e1844-e1844 (2015-08-08)
In the central nervous system (CNS), hyperglycemia leads to neuronal damage and cognitive decline. Recent research has focused on revealing alterations in the brain in hyperglycemia and finding therapeutic solutions for alleviating the hyperglycemia-induced cognitive dysfunction. Adiponectin is a protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service