Skip to Content
Merck
All Photos(1)

Key Documents

EHU110591

Sigma-Aldrich

MISSION® esiRNA

targeting human TDGF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACAGAACCTGCTGCCTGAATGGGGGAACCTGCATGCTGGGGTCCTTTTGTGCCTGCCCTCCCTCCTTCTACGGACGGAACTGTGAGCACGATGTGCGCAAAGAGAACTGTGGGTCTGTGCCCCATGACACCTGGCTGCCCAAGAAGTGTTCCCTGTGTAAATGCTGGCACGGTCAGCTCCGCTGCTTTCCTCAGGCATTTCTACCCGGCTGTGATGGCCTTGTGATGGATGAGCACCTCGTGGCTTCCAGGACTCCAGAACTACCACCGTCTGCACGTACTACCACTTTTATGCTAGTTGGCATCTGCCTTTCTATACAAAGCTACTATTAATCGACATTGACCTATTTCCAGAAATACAATTTTAGATATCATGCAAATTTCATGACCAGTAAAGGCTGCTGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Harpelunde Poulsen et al.
BMC cancer, 20(1), 349-349 (2020-04-25)
Testicular germ cell tumours (TGCTs) are characterised by an overall high cisplatin-sensitivity which has been linked to their continued expression of pluripotency factors. Recently, the Nodal signalling pathway has been implicated in the regulation of pluripotency factor expression in fetal
E Zoni et al.
Oncogene, 36(33), 4739-4749 (2017-04-11)
CRIPTO (CR-1, TDGF1) is a cell surface/secreted oncoprotein actively involved in development and cancer. Here, we report that high expression of CRIPTO correlates with poor survival in stratified risk groups of prostate cancer (PCa) patients. CRIPTO and its signaling partner

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service