Skip to Content
Merck
All Photos(1)

Key Documents

EHU106471

Sigma-Aldrich

MISSION® esiRNA

targeting human GSTO1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Availability
Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Availability
Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGACGCGTCTAGTCCTGAAGGCCAAGGGAATCAGGCATGAAGTCATCAATATCAACCTGAAAAATAAGCCTGAGTGGTTCTTTAAGAAAAATCCCTTTGGTCTGGTGCCAGTTCTGGAAAACAGTCAGGGTCAGCTGATCTACGAGTCTGCCATCACCTGTGAGTACCTGGATGAAGCATACCCAGGGAAGAAGCTGTTGCCGGATGACCCCTATGAGAAAGCTTGCCAGAAGATGATCTTAGAGTTGTTTTCTAAGGTGCCATCCTTGGTAGGAAGCTTTATTAGAAGCCAAAATAAAGAAGACTATGCTGGCCTAAAAGAAGAATTTCGTAAAGAATTTACCAAGCTAGAGGAGGTTCTGACTAATAAGAAGACGACCTTCTTTGGTGGCAATTCTATCTCTATGATTGATTACCTCATCTGGCCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU009201EHU015351EHU056581
product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

GGACGCGTCTAGTCCTGAAGGCCAAGGGAATCAGGCATGAAGTCATCAATATCAACCTGAAAAATAAGCCTGAGTGGTTCTTTAAGAAAAATCCCTTTGGTCTGGTGCCAGTTCTGGAAAACAGTCAGGGTCAGCTGATCTACGAGTCTGCCATCACCTGTGAGTACCTGGATGAAGCATACCCAGGGAAGAAGCTGTTGCCGGATGACCCCTATGAGAAAGCTTGCCAGAAGATGATCTTAGAGTTGTTTTCTAAGGTGCCATCCTTGGTAGGAAGCTTTATTAGAAGCCAAAATAAAGAAGACTATGCTGGCCTAAAAGAAGAATTTCGTAAAGAATTTACCAAGCTAGAGGAGGTTCTGACTAATAAGAAGACGACCTTCTTTGGTGGCAATTCTATCTCTATGATTGATTACCTCATCTGGCCC

esiRNA cDNA target sequence

GGAGACCTCACCCTGTACCAGTCCAATACCATCCTGCGTCACCTGGGCCGCACCCTTGGGCTCTATGGGAAGGACCAGCAGGAGGCAGCCCTGGTGGACATGGTGAATGACGGCGTGGAGGACCTCCGCTGCAAATACATCTCCCTCATCTACACCAACTATGAGGCGGGCAAGGATGACTATGTGAAGGCACTGCCCGGGCAACTGAAGCCTTTTGAGACCCTGCTGTCCCAGAACCAGGGAGGCAAGACCTTCATTGTGGGAGACCAGATCTCCTTCGCTGACTACAACCTGCTGGACTTGCTGCTGATCCATGAGGTCCTAGCCCCTGGCTGCCTGGATGCGTTCCCCCTGCTCTCAGCATATGTGGGGCGCCTCAGTGCCCGGCCCAAGCTCAAGGCCTTCCTGGCCTCCCCTGAGTACGTGAACCTCCC

esiRNA cDNA target sequence

CGGTGTCTGCAGTGATGAAGACCATTCGTATTTTTCAGAAGTTGAATTATATGCTTTTGGCAAAACGTCTTCAGGAGGAGAAGGAGAAAGTTGATAAGCAATACAAATCGGTAACAAAACCAAAGCCACTAAGTCAAGCTGAAATGAGTCCTGCTGCATCTGCAGCCATCAGAAATGATGTCGCAAAGCAACGTGCTGCACCAAAAGTCTCTCCTCATGTTAAGCCTGAACAGAAACAGATGGTGGCCCAGCAGGAATCTATCAGAGAAGGGTTTCAGAAGCGCTGTTTGCCAGTTATGGTACTGAAAGCAAAGAAGCCCTTCACGTTTGAGACCCAAGAAGGCAAGCAGGAGATGTTTCATGCTACAGTGGCTACAGAAAAGGAATTCTTCTTTGTAAAAGTTTTTAATACACTGCTGAAAGATAAATTCATTCCAAAGAGAATAATTATAATAGCAAGATATTATCGGCACAGTGGTTTCTTAGAGGTAAATAGCGCCTCACGTGTG

esiRNA cDNA target sequence

TCCTGTGCCGGTATCAGAATATCTGGAACATCAACCTGCAGTTGCGGCCCAGCCTCATAACAGGGATCATGAAAGACAGTGGAAACAAGCCTCCAGGTCTGCTTCCCCGCAAAGGACTATACATGGCAAATGACTTAAAGCTCCTGAGACACCATCTCCAGATTCCCATCCACTTCCCCAAGGATTTCTTGTCTGTGATGCTTGAAAAAGGAAGTTTGTCTGCCATGCGTTTCCTCACCGCCGTGAACTTGGAGCATCCAGAGATGCTGGAGAAAGCGTCCCGGGAGCTGTGGATGCGCGTCTGGTCAAGGAATGAAGACATCACCGAGCCGCAGAGCATCCTGGCGGCTGCAGAGAAGGCTGGTATGTCTGCAGAACAAGCCCAGGGACTTCTGGAAAAGATCGCAACGCCAAAGGTGAAGAACCAGCTCAAGGAGACCACTGAGG

Ensembl | human accession no.

ENSG00000148834

Ensembl | human accession no.

ENSG00000084207

Ensembl | human accession no.

ENSG00000163568

Ensembl | human accession no.

ENSG00000197448

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kanakaraju Manupati et al.
The FEBS journal, 286(11), 2167-2192 (2019-03-16)
Glutathione S-transferase omega 1 (GSTO1) contributes to the inactivation of a wide range of drug compounds via conjugation to glutathione during phase reactions. Chemotherapy-induced GSTO1 expression in breast cancer cells leads to chemoresistance and promotes metastasis. In search of novel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service