Skip to Content
Merck
All Photos(1)

Key Documents

EHU104281

Sigma-Aldrich

MISSION® esiRNA

targeting human PHB2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTGTCAACGAGGTGCTCAAGAGTGTGGTGGCCAAGTTCAATGCCTCACAGCTGATCACCCAGCGGGCCCAGGTATCCCTGTTGATCCGCCGGGAGCTGACAGAGAGGGCCAAGGACTTCAGCCTCATCCTGGATGATGTGGCCATCACAGAGCTGAGCTTTAGCCGAGAGTACACAGCTGCTGTAGAAGCCAAACAAGTGGCCCAGCAGGAGGCCCAGCGGGCCCAATTCTTGGTAGAAAAAGCAAAGCAGGAACAGCGGCAGAAAATTGTGCAGGCCGAGGGTGAGGCCGAGGCTGCCAAGATGCTTGGAGAAGCACTGAGCAAGAACCCTGGCTACATCAAACTTCGCAAGATTCGAGCAGCCCAGAATATCTCCAAGACGATCGCCACATCACAGAATCGTATCTATCTCACAGCTGACAACCTTGTGCTGAACCTACAGGATGAAAGTTTCACCAGGTACAGTGACAGCCTCATCAAGGGT

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tongyu Zhang et al.
Stroke, 50(4), 978-988 (2019-03-21)
Background and Purpose- Mitoquinone has been reported as a mitochondria-targeting antioxidant to promote mitophagy in various chronic diseases. Here, our aim was to study the role of mitoquinone in mitophagy activation and oxidative stress-induced neuronal death reduction after subarachnoid hemorrhage
Ting Huang et al.
Frontiers in immunology, 11, 569173-569173 (2020-10-30)
Mitophagy has recently been implicated in bacterial infection but the underlying mechanism remains largely unknown. Here, we uncover a role of microRNA-302/367 cluster in regulating mitophagy and its associated host response against bacterial infection. We demonstrate that miR-302/367 cluster expression
Heng-Hsiung Wu et al.
Journal of food and drug analysis, 28(1), 183-194 (2019-12-31)
Membranous nephropathy (MN) is the most common cause of nephrotic syndrome in adults, when not effectively treated. The aim of this study was to discover new targets for the diagnosis and treatment of MN. A reliable mouse model of MN
Cristina Moncunill-Massaguer et al.
Oncotarget, 6(39), 41750-41765 (2015-10-27)
We previously described diaryl trifluorothiazoline compound 1a (hereafter referred to as fluorizoline) as a first-in-class small molecule that induces p53-independent apoptosis in a wide range of tumor cell lines. Fluorizoline directly binds to prohibitin 1 and 2 (PHBs), two proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service