Skip to Content
Merck
All Photos(1)

Key Documents

EHU098571

Sigma-Aldrich

MISSION® esiRNA

targeting human MDM2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCTTCCATCACATTGCAACAGATGTTGGGCCCTTCGTGAGAATTGGCTTCCTGAAGATAAAGGGAAAGATAAAGGGGAAATCTCTGAGAAAGCCAAACTGGAAAACTCAACACAAGCTGAAGAGGGCTTTGATGTTCCTGATTGTAAAAAAACTATAGTGAATGATTCCAGAGAGTCATGTGTTGAGGAAAATGATGATAAAATTACACAAGCTTCACAATCACAAGAAAGTGAAGACTATTCTCAGCCATCAACTTCTAGTAGCATTATTTATAGCAGCCAAGAAGATGTGAAAGAGTTTGAAAGGGAAGAAACCCAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chengtao Sun et al.
OncoTargets and therapy, 13, 10475-10487 (2020-10-30)
Cell-division cycle 20 (CDC20) is overexpressed in a variety of tumor cells and is negatively regulated by wild-type p53 (wtp53). Our previous study uncovered that CDC20 was upregulated and associated with poor outcome in diffuse large B-cell lymphoma (DLBCL) based
Shaneabbas Raza et al.
Molecular and cellular biochemistry, 410(1-2), 187-195 (2015-09-10)
Estrogen is synthesized from cholesterol and high cholesterol levels are suggested to be associated with increased risk of estrogen receptor(ER)-positive breast cancer. The cholesterol metabolite 27-hydroxycholesterol (27-OHC) was recently identified as a selective estrogen receptor modulator (SERM) and may therefore
Kiyohiro Ando et al.
Journal of oncology, 2020, 2752417-2752417 (2020-10-06)
Checkpoint kinase 1 (CHK1) plays a key role in genome surveillance and integrity throughout the cell cycle. Selective inhibitors of CHK1 (CHK1i) are undergoing clinical evaluation for various human malignancies, including neuroblastoma. Recently, we reported that CHK1i, PF-477736, induced a
Bin Zhang et al.
Oncotarget, 8(42), 71894-71910 (2017-10-27)
Growing evidence indicates that 14-3-3ζ and yes-associated protein (YAP) substantially promote tumorigenesis and tumor development. However, the regulatory mechanism underlying these two proteins remains unknown. Herein, we report a new regulatory role of 14-3-3ζ in the phosphorylation of YAP and
Lingxin Meng et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 107, 139-145 (2018-08-08)
Vascular endothelial growth factor (VEGF) signaling promotes angiogenesis by stimulating the migration and proliferation of endothelial cells. The aim of this study was to investigate the expression of Survivin and VEGF receptor 1/2/3 (VEGFR 1/2/3) in esophageal carcinoma tissues (ECTs)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service