Skip to Content
Merck
All Photos(1)

Key Documents

EHU095241

Sigma-Aldrich

MISSION® esiRNA

targeting human FGFR3 (1)

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGTGCAGGTTCCGATGTTATTAGATGTTACAAGTTTATATATATCTATATATATAATTTATTGAGTTTTTACAAGATGTATTTGTTGTAGACTTAACACTTCTTACGCAATGCTTCTAGAGTTTTATAGCCTGGACTGCTACCTTTCAAAGCTTGGAGGGAAGCCGTGAATTCAGTTGGTTCGTTCTGTACTGTTACTGGGCCCTGAGTCTGGGCAGCTGTCCCTTGCTTGCCTGCAGGGCCATGGCTCAGGGTGGTCTCTTCTTGGGGCCCAGTGCATGGTGGCCAGAGGTGTCACCCAAACCG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josine M Quispel-Janssen et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 24(1), 84-94 (2017-10-25)
Purpose: Despite intense research, treatment options for patients with mesothelioma are limited and offer only modest survival advantage. We screened a large panel of compounds in multiple mesothelioma models and correlated sensitivity with a range of molecular features to detect
Xina Xie et al.
Experimental and therapeutic medicine, 18(2), 1226-1234 (2019-07-19)
Fibroblast growth factor receptor 3 (FGFR3) is a high frequency mutant gene in bladder cancer (BCa) and has become a promising therapeutic target due to its involvement in cell proliferation and migration. However, whether and how FGFR3 mutations affects BCa
Evan K Day et al.
Cell reports, 30(10), 3383-3396 (2020-03-12)
SPRY2 is a purported tumor suppressor in certain cancers that promotes tumor growth and resistance to receptor tyrosine kinase inhibitors in glioblastoma. Here, we identify a SPRY2-dependent bypass signaling mechanism in glioblastoma that drives resistance to EGFR and MET inhibition.
Takeshi Okada et al.
Molecular neurobiology, 56(12), 8203-8219 (2019-06-17)
Neuronal apoptosis is a common and critical pathology following subarachnoid hemorrhage (SAH). We investigated the anti-apoptotic property of fibroblast growth factor (FGF)-2 after SAH in rats. A total of 289 rats underwent endovascular perforation to induce SAH or sham operation.
V Tassinari et al.
Cell death & disease, 6, e1688-e1688 (2015-03-15)
Both fibroblast growth factor 9 (Fgf9) and Kit Ligand (Kl) signal through tyrosine kinase receptors, yet they exert opposite effects on meiotic differentiation in postnatal spermatogonia, Fgf9 acting as a meiosis-inhibiting substance and Kl acting as a promoter of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service