Skip to Content
Merck
All Photos(1)

Key Documents

EHU093291

Sigma-Aldrich

MISSION® esiRNA

targeting human IRAK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATCCTCCAGAAAGTCGAGCAGCACCCACCTCCAACCTCGGGCCAGTGTCTTCAGGCTTTACTGGGGACCTGCGAGCTGGCCTAATGTGGTGGCCTGCAAGCCAGGCCATCCCTGGGCGCCACAGACGAGCTCCGAGCCAGGTCAGGCTTCGGAGGCCACAAGCTCAGCCTCAGGCCCAGGCACTGATTGTGGCAGAGGGGCCACTACCCAAGGTCTAGCTAGGCCCAAGACCTAGTTACCCAGACAGTGAGAAGCCCCTGGAAGGCAGAAAAGTTGGGAGCATGGCAGACAGGGAAGGGAAACATTTTCAGGGAAAAGACATGTATCACATGTCTTCAGAAGCAAGTCAGGTTTCATGTAACCGAGTGTCCTCTTGCGTGTCCAAAAGTAGCCCAGGGCTGTAGCACAGGCTTCACAGTGATT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Chen et al.
OncoTargets and therapy, 13, 12787-12796 (2020-12-29)
Interleukin-1 receptor-associated kinase 1 (IRAK1) was shown to contribute to a variety of cancer-related processes. However, the function of IRAK1 in hepatocellular carcinoma (HCC) pathogenesis has not been investigated in detail. IRAK1 expression in HCC was examined by immunohistochemistry, qRT-PCR
Xian Shuang Liu et al.
Molecular neurobiology, 54(1), 227-237 (2016-01-08)
Stroke induces new myelinating oligodendrocytes that are involved in ischemic brain repair. Molecular mechanisms that regulate oligodendrogenesis have not been fully investigated. MicroRNAs (miRNAs) are small non-coding RNA molecules that post-transcriptionally regulate gene expression. MiR-146a has been reported to regulate
Yan Gao et al.
Molecular medicine reports, 14(6), 5685-5692 (2016-11-24)
The present study aimed to reduce the expression of interleukin-1 receptor-associated kinase 1 (IRAK-1) in dendritic cells (DCs) by RNA interference (RNAi). Subsequently, its effects on the expression of costimulatory surface molecules, the release of inflammatory cytokines, and the proliferation of
Hong-Yi Zhang et al.
Journal of pediatric surgery, 55(11), 2308-2316 (2020-04-24)
To investigate the effects of low dose endotoxin on transcriptional activity in intestinal epithelium, and its role in necrotizing enterocolitis (NEC). Lipopolysaccharides (LPS) were injected into the amniotic cavity of pregnant mice under ultrasound guidance. The effects of LPS on
Basak Icli et al.
American journal of physiology. Cell physiology, 318(3), C524-C535 (2020-01-09)
Neoangiogenesis is critical for tissue repair in response to injury such as myocardial ischemia or dermal wound healing. MicroRNAs are small noncoding RNAs and important regulators of angiogenesis under physiological and pathological disease states. Therefore, identification of microRNAs that may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service