Skip to Content
Merck
All Photos(1)

Key Documents

EHU093121

Sigma-Aldrich

MISSION® esiRNA

targeting human CTTN

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTTGGGAAGGAAGGCAGTGCCTGCTCTGCTGTGAGCCGCCAGGAACCCTCCTCCTGTCAATGGGGGTGTAGTATTTTTGCCAAAATATCATGTTCAATTTCAGTAGTTTGATCAGTTGAAGGCTAGAAGTGTGAAGTGCAGATGAGTGTGTGTTCTTCCCCAAGGTCCCCCCACAGCTCCAGGACACCGCTGTCCTGGCATTTGTGGCCACTCACTTTGTAGGAAACTCATCTCCTTCCTGAGGAGCCGGGAGGCTGGACCAGTCCCGTCGTGCAGTCAGGTGGGCGGTGTGTCTTTCCAGAAGGTCACGTGGAAATGTCTCGGGACTTGGGTCCCGGAGTGCCCGTGAAGCGTGTTTTTGCTCCTGAGGTGCATTTTCTCATCATCCTTGCTTTACCACAATGAGCAATGAGGTCGGGTTTTA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojian Zhang et al.
Oncotarget, 8(1), 1541-1554 (2016-12-03)
Cortactin (CTTN) is overexpressed in various tumors, including head and neck squamous cell carcinoma and colorectal cancer (CRC), and can serve as a biomarker of cancer metastasis. We observed that CTTN promotes cancer cell proliferation in vitro and increases CRC
Rachel J Watkins et al.
The Journal of clinical endocrinology and metabolism, 101(12), 4551-4563 (2016-09-08)
Metastatic disease is responsible for the majority of endocrine cancer deaths. New therapeutic targets are urgently needed to improve patient survival rates. The proto-oncogene PTTG1-binding factor (PBF/PTTG1IP) is overexpressed in multiple endocrine cancers and circumstantially associated with tumor aggressiveness. This
Steven M Markwell et al.
Molecular cancer research : MCR, 17(4), 987-1001 (2019-01-06)
Malregulation of the actin cytoskeleton enhances tumor cell motility and invasion. The actin-binding protein cortactin facilitates branched actin network formation through activation of the actin-related protein (Arp) 2/3 complex. Increased cortactin expression due to gene amplification is observed in head
Xuan Liang et al.
Nature communications, 8(1), 790-790 (2017-10-07)
Contractile adherens junctions support cell-cell adhesion, epithelial integrity, and morphogenesis. Much effort has been devoted to understanding how contractility is established; however, less is known about whether contractility can be actively downregulated at junctions nor what function this might serve.
Dominik Horn et al.
Head & neck, 40(12), 2685-2694 (2018-11-21)
Cortactin (CTTN) is located on chromosome 11q13 and is associated with invasiveness in various cancer entities. CTTN protein expression could be a prognosticator of oral squamous cell carcinoma (OSCC) in terms of recurrence and survival. CTTN-dependent invasion was performed using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service