Skip to Content
Merck
All Photos(1)

Key Documents

EHU092071

Sigma-Aldrich

MISSION® esiRNA

targeting human FTO

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTGAAGATTGGCCTCTTTCCTTTCTCTAAGACAAACCTAAGTAAAAGCCTGAGCTTTGAGTCCTATGCTCAGCACACGGGAAGGAGATGTTAATAATTAAAATAAAGTTGATATCCTGTCTTTAGGGAGTTCCCTTGATCTCTTGAAAGAGACACAGCCCCATTTACATTATTTCGTGGATTTCACCAGCATAGTATAGTTTTTTTCTGTAAGTCCCTCATTCTTATGTAATAACAGGTGGAACTGAGGTTTGAAGAACCTCAGTGGCCCATCCTGATGACATTGGAGACTCAAAGAGACAAGAGAGAGTAGGGTTTAAAACCTGAGCTTTAAGACTCCCACTAGCTTCGTGTCCTTTGGCATGTTAACGTGCCTCAGTTTCCTCATCTGTATAATGGGGATATATGAAAGGCACCAGTCCTAAGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Xu et al.
Oncology reports, 38(4), 2285-2292 (2017-08-30)
Fat mass and obesity associated (FTO) is a protein-coding gene. FTO gene is an obesity related gene, also known as the obesity gene. It has been reported previously that FTO is associated with a variety of malignant cancers, such as
Ziqi Ye et al.
Oncology letters, 20(2), 1409-1417 (2020-07-30)
Liver cancer is the fourth leading cause of cancer-associated mortality worldwide. Statistics indicate that the incidence of liver cancer has been increasing and that its prognosis remains poor. Fat mass and obesity-associated protein (FTO) is a demethylase that is involved
Ruifan Wu et al.
Biochimica et biophysica acta. Gene regulatory mechanisms, 1862(8), 796-806 (2019-07-12)
N6-methyladenosine (m6A), the most abundant internal mRNA modification in eukaryotes, plays a vital role in regulating adipogenesis. However, its underlying mechanism remains largely unknown. Here, we reveal that deletion of m6A demethylase FTO in porcine and mouse preadipocytes inhibits adipogenesis
Dong Ma et al.
Frontiers in cardiovascular medicine, 7, 592550-592550 (2020-12-18)
Background: Aortic dissecting aneurysm (ADA) represents an aortic remodeling disease with a high mortality rate. Fat mass and obesity-associated protein (FTO) exerts RNA demethylation function to regulate gene expression related to stem cell differentiation, DNA damage repair, and tumorigenesis, but
Chenyue Ding et al.
Journal of cellular physiology, 233(9), 7055-7066 (2018-02-01)
The N6-methyladenosine (m6A) modification plays a central role in epigenetic regulation of the mammalian transcriptome. m6A can be demethylated by the fat mass- and obesity-associated (FTO) protein and the α-ketoglutarate-dependent dioxygenase alkB homolog 5 (ALKBH5) protein. Much less is known

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service