Skip to Content
Merck
All Photos(1)

Key Documents

EHU086561

Sigma-Aldrich

MISSION® esiRNA

targeting human MBNL1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGCCGAACATCTGACTAGCCACAAGTATGTTACCCAGATGTAGAATTTTCATCACTAAACAATCATGCTAAAGAGGAAAGGACAGTGTGCTTGGTTAGAGTAAAGGACGAGGTCATTAGCCATATTGTATATATCGTCAAGCAACACACACAAAAGTTCCTCAGCCACAAGACATCCACATATTGCATGTTAACCAGAAGAAAAGACAACATTTTCCGGAAATCCACTGCACACTGTTGCCTATACACTTTGTACATTTAATTGATATTTGTGCTGAGGTGATATTCCTGTCTAAAAGAACAACATTGTCTTTCTTTTCTAGCACAGAGTTATGCATTCAAAGATGCATACCTAGTTAGTTTCCTATATATTCATGCCATCTTGAAAAGACAGACTATGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Svetlana S Itskovich et al.
Nature communications, 11(1), 2369-2369 (2020-05-14)
Despite growing awareness of the biologic features underlying MLL-rearranged leukemia, targeted therapies for this leukemia have remained elusive and clinical outcomes remain dismal. MBNL1, a protein involved in alternative splicing, is consistently overexpressed in MLL-rearranged leukemias. We found that MBNL1
Łukasz J Sznajder et al.
Nature communications, 11(1), 2022-2022 (2020-04-26)
The thymus is a primary lymphoid organ that plays an essential role in T lymphocyte maturation and selection during development of one arm of the mammalian adaptive immune response. Although transcriptional mechanisms have been well documented in thymocyte development, co-/post-transcriptional
Sandra Fischer et al.
RNA (New York, N.Y.), 26(5), 648-663 (2020-03-05)
Hypoxia is a hallmark of solid cancers, supporting proliferation, angiogenesis, and escape from apoptosis. There is still limited understanding of how cancer cells adapt to hypoxic conditions and survive. We analyzed transcriptome changes of human lung and breast cancer cells
Hongfei Liu et al.
Nucleic acids research, 46(12), 6069-6086 (2018-05-18)
We report the detailed transcriptomic profiles of human innate myeloid cells using RNA sequencing. Monocytes migrate from blood into infected or wounded tissue to differentiate into macrophages, and control inflammation via phagocytosis or cytokine secretion. We differentiated culture primary monocytes
Xin Sun et al.
Scientific reports, 5, 12521-12521 (2015-07-29)
Huntington's disease (HD) is caused by a CAG repeat expansion in the huntingtin (HTT) gene. Recent evidence suggests that HD is a consequence of multimodal, non-mutually exclusive mechanisms of pathogenesis that involve both HTT protein- and HTT RNA-triggered mechanisms. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service