Skip to Content
Merck
All Photos(1)

Key Documents

EHU078971

Sigma-Aldrich

MISSION® esiRNA

targeting human TFRC

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCCAGCAAAGTTGAGAAACTCACTTTAGACAATGCTGCTTTCCCTTTCCTTGCATATTCTGGAATCCCAGCAGTTTCTTTCTGTTTTTGCGAGGACACAGATTATCCTTATTTGGGTACCACCATGGACACCTATAAGGAACTGATTGAGAGGATTCCTGAGTTGAACAAAGTGGCACGAGCAGCTGCAGAGGTCGCTGGTCAGTTCGTGATTAAACTAACCCATGATGTTGAATTGAACCTGGACTATGAGAGGTACAACAGCCAACTGCTTTCATTTGTGAGGGATCTGAACCAATACAGAGCAGACATAAAGGAAATGGGCCTGAGTTTACAGTGGCTGTATTCTGCTCGTGGAGACTTCTTCCGTGCTACTTCCAGACTAACAACAGATTTCGGGAATGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yihe Wu et al.
Thoracic cancer, 9(2), 253-261 (2017-12-30)
Transferrin receptor (TfR) is expressed in most lung cancers and is an indicator of poor prognosis in certain groups of patients. In this study, we blocked cell surface TfR to inhibit lung adenocarcinoma (LAC) cell growth in vitro and investigated
Hongwei Su et al.
Molecular cancer, 18(1), 27-27 (2019-02-21)
Circular RNA (circRNA) represents a broad and diverse endogenous RNAs that can regulate gene expression in cancer. However, the regulation and function of bladder cancer (BC) circRNAs remain largely unknown. Here we generated circRNA microarray data from three BC tissues
Tian Tang et al.
Singapore medical journal, 62(2), 96-103 (2019-11-05)
Dihydroartemisinin (DHA) is a first-line antimalarial drug with relatively low toxicity. DHA has been speculated to possess a broad-spectrum antitumour effect. However, the potential value of DHA for the treatment of endometrial carcinoma or cervical cancer is unclear. We used
Markus von Nickisch-Rosenegk et al.
Journal of nanobiotechnology, 10, 1-1 (2012-01-10)
The need to functionalize cell membranes in a directed way for specific applications as single cell arrays or to force close cell-to-cell contact for artificial intercellular interaction and/or induction concerning stem cell manipulation or in general to have a tool
Julie Mazzolini et al.
The Biological bulletin, 231(1), 40-60 (2016-09-18)
Particles present in diesel exhaust have been proposed as a significant contributor to the development of acute and chronic lung diseases, including respiratory infection and allergic asthma. Nanoceria (CeO2 nanoparticles) are used to increase fuel efficiency in internal combustion engines

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service