Skip to Content
Merck
All Photos(1)

Key Documents

EHU078751

Sigma-Aldrich

MISSION® esiRNA

targeting human ERBB2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCATCTGCACCATTGATGTCTACATGATCATGGTCAAATGTTGGATGATTGACTCTGAATGTCGGCCAAGATTCCGGGAGTTGGTGTCTGAATTCTCCCGCATGGCCAGGGACCCCCAGCGCTTTGTGGTCATCCAGAATGAGGACTTGGGCCCAGCCAGTCCCTTGGACAGCACCTTCTACCGCTCACTGCTGGAGGACGATGACATGGGGGACCTGGTGGATGCTGAGGAGTATCTGGTACCCCAGCAGGGCTTCTTCTGTCCAGACCCTGCCCCGGGCGCTGGGGGCATGGTCCACCACAGGCACCGCAGCTCATCTACCAGGAGTGGCGGTGGGGACCTGACACTAGGGCTGGAGCCCTCTGAAGAGGAGGCCCCCAGGTCTCCACTGGCACCCTCCGAAGGGGCTGGCTCCGATGTATTTGATGGTGACCTGGGAAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiia Honkanen et al.
International journal of oncology, 51(2), 599-606 (2017-06-29)
We have previously shown that cancer stem-like cells (CSLCs) can mediate therapy resistance in ALK translocated lung cancers. HER2 has been linked to CSLCs in breast cancers and, therefore, we wanted to assess whether HER2 has a role in CSLCs
Nehal Gupta et al.
Scientific reports, 9(1), 5066-5066 (2019-03-27)
Paclitaxel is a first line chemotherapeutic agent for the patients with metastatic breast cancer. But inherited or acquired resistance to paclitaxel leads to poor response rates in a majority of these patients. To identify mechanisms of paclitaxel resistance, we developed
T-T Yu et al.
European review for medical and pharmacological sciences, 24(10), 5267-5280 (2020-06-05)
To explore possible mechanism of ERBB2 gene expression silencing mediating mitogen-activated protein kinase 1/mitogen-activated protein kinase 3 (MAPK1/MAPK3) signaling pathway on proliferation, migration, and invasion of ovarian cancer cells. A total of 240 cancer specimens were collected in patients with
Tong Shu et al.
Cancer letters, 411, 65-73 (2017-10-11)
Resistance to platinum-based chemotherapy is a major cause of treatment failure in patients with epithelial ovarian cancer and predicts a poor prognosis. Previously, we found that HECTD3 confers cancer cell resistance to apoptosis. However, the significance of HECTD3 expression in
Yiseul Choi et al.
World journal of gastroenterology, 22(41), 9141-9153 (2016-11-30)
To investigated the relationships between HER2, c-Jun N-terminal kinase (JNK) and protein kinase B (AKT) with respect to metastatic potential of HER2-positive gastric cancer (GC) cells. Immunohistochemistry was performed on tissue array slides containing 423 human GC specimens. Using HER2-positve

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service