Skip to Content
Merck
All Photos(1)

Documents

EHU075891

Sigma-Aldrich

MISSION® esiRNA

targeting human SOD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTTGGCCAAGGGAGATGTTACAGCCCAGATAGCTCTTCAGCCTGCACTGAAGTTCAATGGTGGTGGTCATATCAATCATAGCATTTTCTGGACAAACCTCAGCCCTAACGGTGGTGGAGAACCCAAAGGGGAGTTGCTGGAAGCCATCAAACGTGACTTTGGTTCCTTTGACAAGTTTAAGGAGAAGCTGACGGCTGCATCTGTTGGTGTCCAAGGCTCAGGTTGGGGTTGGCTTGGTTTCAATAAGGAACGGGGACACTTACAAATTGCTGCTTGTCCAAATCAGGATCCACTGCAAGGAACAACAGGCCTTATTCCACTGCTGGGGATTGATGTGTGGGAGCACGCTTACTACCTTCAGTATAAAAATGTCAGGCCTGATTATCTAAAAGCTATTTGGAATGTAATCAACTGGGAGAATGTAACTGAAAGATACATGGCTTGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miyoung Lee et al.
Antioxidants (Basel, Switzerland), 9(1) (2020-01-17)
Umbilical cord blood-derived mesenchymal stem cells (UCB-MSCs) are accessible, available in abundance, and have been shown to be a promising source that can regenerate cartilage in patients with osteoarthritis or other orthopedic diseases. Recently, a three-dimensional (3D) cell culture system
Serena Sagliocchi et al.
Redox biology, 24, 101228-101228 (2019-06-04)
Thyroid hormone (TH) is a key metabolic regulator that acts by coordinating short- and long-term energy needs. Accordingly, significant metabolic changes are observed depending on thyroid status. Although it is established that hyperthyroidism augments basal energy consumption, thus resulting in
Veronica Nobile et al.
Human genetics, 139(2), 227-245 (2020-01-11)
Fragile X-related disorders are due to a dynamic mutation of the CGG repeat at the 5' UTR of the FMR1 gene, coding for the RNA-binding protein FMRP. As the CGG sequence expands from premutation (PM, 56-200 CGGs) to full mutation
You Dong Liu et al.
International journal of cancer, 144(12), 3056-3069 (2018-12-12)
Toll-like receptors (TLRs) play critical roles in host defense after recognition of conserved microbial- and host-derived components, and their dysregulation is a common feature of various inflammation-associated cancers, including gastric cancer (GC). Despite the recent recognition that metabolic reprogramming is
Brianna M Young et al.
Molecular therapy. Methods & clinical development, 14, 113-125 (2019-07-25)
Age-related macular degeneration (AMD) has been linked to oxidative damage and para-inflammation, an activation of inflammasome signaling in the retinal pigment epithelium (RPE) and the underlying choriocapillaris. Herein, we tested the efficacy of a gene-delivered caspase-1 inhibitor in controlling the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service