Skip to Content
Merck
All Photos(1)

Key Documents

EHU075051

Sigma-Aldrich

MISSION® esiRNA

targeting human EPHA2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGGACCTGATGCAGAACATCATGAATGACATGCCGATCTACATGTACTCCGTGTGCAACGTGATGTCTGGCGACCAGGACAACTGGCTCCGCACCAACTGGGTGTACCGAGGAGAGGCTGAGCGTATCTTCATTGAGCTCAAGTTTACTGTACGTGACTGCAACAGCTTCCCTGGTGGCGCCAGCTCCTGCAAGGAGACTTTCAACCTCTACTATGCCGAGTCGGACCTGGACTACGGCACCAACTTCCAGAAGCGCCTGTTCACCAAGATTGACACCATTGCGCCCGATGAGATCACCGTCAGCAGCGACTTCGAGGCACGCCACGTGAAGCTGAACGTGGAGGAGCGCTCCGTGGGGCCGCTCACCCGCAAAGGCTTCTACCTGGCCTTCCAGGATATCGGTGCCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiaoli Wu et al.
Biochemical and biophysical research communications, 503(4), 2436-2442 (2018-07-04)
MiR-124-3p and EphA2 are aberrantly expressed in glioma tissue specimens. In the present study, we firstly investigated that miR-124-3p inhibits EphA2 expression mediated by binding its 3'-UTR to regulate the progression of human glioma. The U87MG and LN229 cells were transfected
Changhao Huang et al.
International journal of cancer, 146(7), 1937-1949 (2019-08-04)
Yes-associated protein (YAP) is a transcriptional coactivator that promotes cell proliferation, stem cell maintenance and tissue homeostasis. The YAP activity is primarily regulated through an inhibitory phosphorylation by the serine/threonine kinases of Hippo pathway. Here, we show that receptor tyrosine
Hee Sung Kim et al.
Scientific reports, 9(1), 3414-3414 (2019-03-06)
Genetically deregulated tumor cells generate vascular channels by vasculogenic mimicry (VM) that is independent of endothelial blood vessels. The morphological characteristics of VM and the role of EphA2 in the formation of VM were evaluated in 144 clinical samples of
Maleeha A Qazi et al.
Cancer research, 78(17), 5023-5037 (2018-06-28)
Glioblastoma (GBM) carries a dismal prognosis and inevitably relapses despite aggressive therapy. Many members of the Eph receptor tyrosine kinase (EphR) family are expressed by GBM stem cells (GSC), which have been implicated in resistance to GBM therapy. In this
Xin Zhang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(4), 5495-5509 (2019-01-23)
The balance of myogenic and adipogenic differentiation is crucial for skeletal muscle homeostasis. Given the vital role of membrane proteins (MBPs) in cell signal perception, membrane proteomics was conducted to delineate mechanisms regulating differentiation of adipogenic and myogenic precursors in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service