Skip to Content
Merck
All Photos(1)

Key Documents

EHU072741

Sigma-Aldrich

MISSION® esiRNA

targeting human RORA

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CGTGCCTTTGACTCTCAGAACAACACCGTGTACTTTGATGGGAAGTATGCCAGCCCCGACGTCTTCAAATCCTTAGGTTGTGAAGACTTTATTAGCTTTGTGTTTGAATTTGGAAAGAGTTTATGTTCTATGCACCTGACTGAAGATGAAATTGCATTATTTTCTGCATTTGTACTGATGTCAGCAGATCGCTCATGGCTGCAAGAAAAGGTAAAAATTGAAAAACTGCAACAGAAAATTCAGCTAGCTCTTCAACACGTCCTACAGAAGAATCACCGAGAAGATGGAATACTAACAAAGTTAATATGCAAGGTGTCTACCTTAAGAGCCTTATGTGGACGACATACAGAAAAGCTAATGGCATTTAAAGCAATATACCCAGACATTGTGCGACTTCATTTTCCTCCATTATACAAGGAGTTGTTCACTTCAGAATTTGAGCCAGCAATGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongyu Li et al.
Journal of cellular physiology, 233(1), 641-650 (2017-03-24)
Low O2 pressures present in the microenvironment of epidermis control keratinocyte differentiation and epidermal barrier function through hypoxia inducible factors (HIFs) dependent gene expression. This study focuses on investigating relations of the retinoic acid receptor-related orphan receptor alpha (RORα) to
Hao Hu et al.
The Biochemical journal, 477(18), 3583-3598 (2020-07-21)
Estrogen sulfotransferase (SULT1E1) metabolically inactivates estrogen and SULT1E1 expression is tightly regulated by multiple nuclear receptors. Human fetal, but not adult, livers express appreciable amounts of SULT1E1 protein, which is mimicked in human hepatoma-derived HepG2 cells cultured in high glucose
Juho Heliste et al.
BMC cardiovascular disorders, 18(1), 196-196 (2018-10-22)
Receptor tyrosine kinases (RTK) are potential targets for the treatment of ischemic heart disease. The human RTK family consists of 55 members, most of which have not yet been characterized for expression or activity in the ischemic heart. RTK gene
Dongli Liu et al.
Scientific reports, 10(1), 13906-13906 (2020-08-19)
ROR1 and ROR2 are receptor tyrosine kinases with altered expression in a range of cancers. Silencing ROR1 or ROR2 in different tumour types has been shown to inhibit proliferation and decrease metastatic potential. The aim of this study was to
Fernanda Faião-Flores et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 25(18), 5686-5701 (2019-06-23)
The clinical use of MEK inhibitors in uveal melanoma is limited by the rapid acquisition of resistance. This study has used multiomics approaches and drug screens to identify the pan-HDAC inhibitor panobinostat as an effective strategy to limit MEK inhibitor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service