Skip to Content
Merck
All Photos(2)

Key Documents

EHU070511

Sigma-Aldrich

MISSION® esiRNA

targeting human CDK4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GAAACTCTGAAGCCGACCAGTTGGGCAAAATCTTTGACCTGATTGGGCTGCCTCCAGAGGATGACTGGCCTCGAGATGTATCCCTGCCCCGTGGAGCCTTTCCCCCCAGAGGGCCCCGCCCAGTGCAGTCGGTGGTACCTGAGATGGAGGAGTCGGGAGCACAGCTGCTGCTGGAAATGCTGACTTTTAACCCACACAAGCGAATCTCTGCCTTTCGAGCTCTGCAGCACTCTTATCTACATAAGGATGAAGGTAATCCGGAGTGAGCAATGGAGTGGCTGCCATGGAAGGAAGAAAAGCTGCCATTTCCCTTCTGGACACTGAGAGGGCAATCTTTGCCTTTATCTCTGAGGCTATGGAGGGTCCTCCTCCATCTTTCTACAGAGATTACTTTGCTGCCTTAATGACATTCCCCTCCCACCTCTCCTTTTGAGGCTTCTCCTTCTCCTTCCCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjing Shang et al.
EBioMedicine, 53, 102672-102672 (2020-03-03)
Abnormal expression of the orphan nuclear receptor Nurr1 is a critical factor in the etiology of multiple cancers. However, its potential role in gastric cancer (GC) remains elusive. In this study, we have demonstrated that the expression of Nurr1 was
Amriti R Lulla et al.
Cancer research, 77(24), 6902-6913 (2017-10-25)
CDK4/6 targeting is a promising therapeutic strategy under development for various tumor types. In this study, we used computational methods and The Cancer Genome Atlas dataset analysis to identify novel miRNAs that target CDK4/6 and exhibit potential for therapeutic development
Liam Cornell et al.
Cell reports, 26(10), 2667-2680 (2019-03-07)
CDK4/6 inhibition is now part of the standard armamentarium for patients with estrogen receptor-positive (ER+) breast cancer, so that defining mechanisms of resistance is a pressing issue. Here, we identify increased CDK6 expression as a key determinant of acquired resistance
Nikhlesh K Singh et al.
The Journal of biological chemistry, 292(34), 14080-14091 (2017-06-29)
Although the involvement of Rho proteins in the pathogenesis of vascular diseases is well studied, little is known about the role of their upstream regulators, the Rho guanine nucleotide exchange factors (RhoGEFs). Here, we sought to identify the RhoGEFs involved
Smruthi Vijayaraghavan et al.
Nature communications, 8, 15916-15916 (2017-06-28)
Deregulation of the cell cycle machinery is a hallmark of cancer. While CDK4/6 inhibitors are FDA approved (palbociclib) for treating advanced estrogen receptor-positive breast cancer, two major clinical challenges remain: (i) adverse events leading to therapy discontinuation and (ii) lack

Articles

Quantitative and qualitative western blotting to validate knockdown by esiRNA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service