Skip to Content
Merck
All Photos(1)

Key Documents

EHU068681

Sigma-Aldrich

MISSION® esiRNA

targeting human PTK2B

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGGGAGGTCTATGAAGGTGTCTACACAAATCACAAAGGGGAGAAAATCAATGTAGCTGTCAAGACCTGCAAGAAAGACTGCACTCTGGACAACAAGGAGAAGTTCATGAGCGAGGCAGTGATCATGAAGAACCTCGACCACCCGCACATCGTGAAGCTGATCGGCATCATTGAAGAGGAGCCCACCTGGATCATCATGGAATTGTATCCCTATGGGGAGCTGGGCCACTACCTGGAGCGGAACAAGAACTCCCTGAAGGTGCTCACCCTCGTGCTGTACTCACTGCAGATATGCAAAGCCATGGCCTACCTGGAGAGCATCAACTGCGTGCACAGGGACATTGCTGTCCGGAACATCCTGGTGGCCTCCCCTGAGTGTGTGAAGCTGGGGGACTTTGGTCTTTCCCGGTACA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chien-Chung Yang et al.
International journal of molecular sciences, 21(1) (2020-01-08)
Neuroinflammation is a landmark of neuroinflammatory and neurodegenerative diseases. Matrix metalloproteinase (MMP)-9, one member of MMPs, has been shown to contribute to the pathology of these brain diseases. Several experimental models have demonstrated that lipopolysaccharide (LPS) exerts a pathological role
Jialin Qian et al.
Oncology letters, 11(3), 1738-1744 (2016-03-22)
Lung cancer, specifically non-small cell lung cancer (NSCLC), is the leading cause of cancer-associated mortality in the world. In previous years, almost no significant advancements have been made towards the molecular characterization of NSCLC, which highlights the requirement for novel
Chih-Chung Lin et al.
Frontiers in molecular neuroscience, 10, 387-387 (2017-12-07)
Neurodegenerative disorders and brain damage are initiated by excessive production of reactive oxygen species (ROS), which leads to tissue injury, cellular death and inflammation. In cellular anti-oxidant systems, heme oxygenase-1 (HO-1) is an oxidative-sensor protein induced by ROS generation or
Leny Jose et al.
Journal of virology, 94(20) (2020-07-31)
The human papillomavirus (HPV) E2 protein is a key regulator of viral transcription and replication. In this study, we demonstrate that the nonreceptor tyrosine kinase Pyk2 phosphorylates tyrosine 131 in the E2 transactivation domain. Both depletion of Pyk2 and treatment
Pradip Das et al.
Journal of immunology (Baltimore, Md. : 1950), 206(1), 181-192 (2020-12-06)
MCP-1-induced monocyte chemotaxis is a crucial event in inflammation and atherogenesis. Identifying the important signal transduction pathways that control monocyte chemotaxis can unravel potential targets for preventive therapies in inflammatory disease conditions. Previous studies have shown that the focal adhesion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service