Skip to Content
Merck
All Photos(1)

Key Documents

EHU062331

Sigma-Aldrich

MISSION® esiRNA

targeting human TRAF3IP2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCAGAAGAATTGCGGAAAGTCTTTATCACTTATTCGATGGACACAGCTATGGAGGTGGTGAAATTCGTGAACTTTTTGTTGGTAAATGGCTTCCAAACTGCAATTGACATATTTGAGGATAGAATCCGAGGCATTGATATCATTAAATGGATGGAGCGCTACCTTAGGGATAAGACCGTGATGATAATCGTAGCAATCAGCCCCAAATACAAACAGGACGTGGAAGGCGCTGAGTCGCAGCTGGACGAGGATGAGCATGGCTTACATACTAAGTACATTCATCGAATGATGCAGATTGAGTTCATAAAACAAGGAAGCATGAATTTCAGATTCATCCCTGTGCTCTTCCCAAATGCTAAGAAGGAGCATGTGCCCACCTGGCTTCAGAACACTCATGTCTACAGCTGGCCCAAGAAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitin A Das et al.
Journal of molecular and cellular cardiology, 121, 107-123 (2018-07-10)
Persistent inflammation promotes development and progression of heart failure (HF). TWEAK (TNF-Related WEAK Inducer Of Apoptosis), a NF-κB- and/or AP-1-responsive proinflammatory cytokine that signals via TWEAK receptor (TWEAKR), is expressed at high levels in human and preclinical models of HF.
Hongxue Sun et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 48(2), 528-539 (2018-07-19)
This study investigated the role of the microRNA miR-298 and its target Act1 in ischemic stroke. Cell viability was assessed with the 3-(4,5-dimethythiazol-2- yl)-2,5-diphenyl tetrazolium bromide assay. Apoptotic cells were detected by flow cytometry, and mRNA and protein expression were
Hyo Jeong Kim et al.
Biochemical and biophysical research communications, 524(4), 1044-1050 (2020-02-19)
Bone homeostasis is maintained by concerted actions of bone-forming osteoblasts and bone-resorbing osteoclasts. A wide range of evidence indicates that a proinflammatory cytokine IL-17 promotes osteoclastogenesis. However, the role of IL-17 in osteoblasts is less well-understood. In the current study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service