Skip to Content
Merck
All Photos(1)

Documents

EHU059261

Sigma-Aldrich

MISSION® esiRNA

targeting human TFEB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCGGCAGAAGAAAGACAATCACAACTTAATTGAAAGGAGACGAAGGTTCAACATCAATGACCGCATCAAGGAGTTGGGAATGCTGATCCCCAAGGCCAATGACCTGGACGTGCGCTGGAACAAGGGCACCATCCTCAAGGCCTCTGTGGATTACATCCGGAGGATGCAGAAGGACCTGCAAAAGTCCAGGGAGCTGGAGAACCACTCTCGCCGCCTGGAGATGACCAACAAGCAGCTCTGGCTCCGTATCCAGGAGCTGGAGATGCAGGCTCGAGTGCACGGCCTCCCTACCACCTCCCCGTCCGGCATGAACATGGCTGAGCTGGCCCAGCAGGTGGTGAAGCAGGAGCTGCCTAGCGAAGAGGGCCCAGGGGAGGCCCTGATGCTGGGGGCTGAGGTCCCTGACCCTGAGCCACTGCCAGCTCTGCCCCCGCAAGCCCCGCTGCCCCTGCCCACCCAGCCACCATCCCCATTCCATCACCTGGACTTCAGCCAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sijie Tan et al.
Scientific reports, 9(1), 727-727 (2019-01-27)
Mitochondrial dysfunction underscores aging and diseases. Mitophagy (mitochondria + autophagy) is a quality control pathway that preserves mitochondrial health by targeting damaged mitochondria for autophagic degradation. Hence, molecules or compounds that can augment mitophagy are therapeutic candidates to mitigate mitochondrial-related diseases. However
Valeria Crippa et al.
Scientific reports, 6, 22827-22827 (2016-03-11)
Neurodegenerative diseases (NDs) are often associated with the presence of misfolded protein inclusions. The chaperone HSPB8 is upregulated in mice, the human brain and muscle structures affected during NDs progression. HSPB8 exerts a potent pro-degradative activity on several misfolded proteins
Samuel Peña-Llopis et al.
The EMBO journal, 30(16), 3242-3258 (2011-08-02)
Mammalian target of rapamycin (mTOR) complex 1 (mTORC1) is an important, highly conserved, regulator of cell growth. Ancient among the signals that regulate mTORC1 are nutrients. Amino acids direct mTORC1 to the surface of the late endosome/lysosome, where mTORC1 becomes
Xingchen Zhao et al.
The Journal of pathology, 245(2), 235-248 (2018-03-24)
Insufficient autophagy in podocytes is related to podocyte injury in diabetic nephropathy (DN). Advanced glycation end-products (AGEs) are major factors of podocyte injury in DN. However, the role and mechanism of AGEs in autophagic dysfunction remain unknown. We investigated autophagic
Hong-Ying Chu et al.
Cancer letters, 423, 105-112 (2018-03-11)
The role of autophagy in tongue squamous cell carcinoma (TSCC) cisplatin resistance is unclear. We aimed to identify a possible synergistic effect of autophagy inhibitors and cisplatin in TSCC cells and explore the underlying mechanism. Our results indicate that autophagic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service