Skip to Content
Merck
All Photos(1)

Key Documents

EHU044171

Sigma-Aldrich

MISSION® esiRNA

targeting human MAPK1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCGCTTCAGACATGAGAACATCATTGGAATCAATGACATTATTCGAGCACCAACCATCGAGCAAATGAAAGATGTATATATAGTACAGGACCTCATGGAAACAGATCTTTACAAGCTCTTGAAGACACAACACCTCAGCAATGACCATATCTGCTATTTTCTCTACCAGATCCTCAGAGGGTTAAAATATATCCATTCAGCTAACGTTCTGCACCGTGACCTCAAGCCTTCCAACCTGCTGCTCAACACCACCTGTGATCTCAAGATCTGTGACTTTGGCCTGGCCCGTGTTGCAGATCCAGACCATGATCACACAGGGTTCCTGACAGAATATGTGGCCACACGTTGGTACAGGGCTCCAGAAATTATGTTGAATTCCAAGGGCTACACCAAGTCCATTGATATTTGGTCTGTAGGCTGCATTCTGGCAGAAATGCTTTCTAACAGGCCCATCTTTCCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeo Dochi et al.
The Journal of general virology, 95(Pt 5), 1156-1166 (2014-02-11)
We reported previously that Pin1 facilitates human immunodeficiency virus type 1 (HIV-1) uncoating by interacting with the capsid core through the phosphorylated Ser(16)-Pro(17) motif. However, the specific kinase responsible for Ser(16) phosphorylation has remained unknown. Here, we showed that virion-associated
Li-Li Xu et al.
Oxidative medicine and cellular longevity, 2018, 3271617-3271617 (2018-06-12)
Ulcerative colitis (UC) is a common inflammatory bowel disease that can destroy the integrity of the colon and increase the risk of colorectal cancer. Oxidative stress is one of the critical pathogenic factors for UC, further impairing the entire affected
Susanne Ulm et al.
Journal of molecular and cellular cardiology, 72, 104-116 (2014-03-19)
Mitogen-activated protein kinases (MAPKs) are involved in the regulation of cardiac hypertrophy and myocyte survival. Extracellular signal regulated protein kinase 1 and 2 (ERK1/2) are key components in the MAPK signaling pathways. Dysfunction of ERK1/2 in congenital heart diseases (Noonan
Jian Zhang et al.
Experimental & molecular medicine, 49(9), e379-e379 (2017-09-25)
The objective of this study was to investigate the regulatory effects of TGF-β1 on CCL3/4 expression and inflammation-related pain during intervertebral disc degeneration (IVDD). TGF-β1 and CCL3/4 expression patterns in different degenerative human nucleus pulposus (NP) tissues were measured by
Takeshi Koujima et al.
Molecular therapy oncolytics, 17, 107-117 (2020-04-24)
Pancreatic ductal adenocarcinoma (PDAC) cells have an exceptional ability to invade nerves through pronounced crosstalk between nerves and cancer cells; however, the mechanism of PDAC cell invasion remains to be elucidated. Here, we demonstrate the therapeutic potential of telomerase-specific oncolytic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service