Skip to Content
Merck
All Photos(1)

Key Documents

EHU042421

Sigma-Aldrich

MISSION® esiRNA

targeting human PIK3C3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCAGCCAAGCATTGTTGAAGGGTGATAAGTCTGTCAGAGTTATGCGTTCTTTGCTGGCTGCACAACAGACATTTGTAGATCGGTTGGTGCATCTAATGAAGGCAGTACAACGCGAAAGTGGAAATCGTAAGAAAAAGAATGAGAGACTACAGGCATTGCTTGGAGATAATGAAAAGATGAATTTGTCAGATGTGGAACTTATCCCGTTGCCTTTAGAACCCCAAGTGAAAATTAGAGGAATAATTCCGGAAACAGCTACACTGTTTAAAAGTGCCCTTATGCCTGCACAGTTGTTTTTTAAGACGGAAGATGGAGGCAAATATCCAGTTATATTTAAGCATGGAGATGATTTACGTCAAGATCAACTTATTCTTCAAATCATTTCACTCATGGACAAGCTGTTACGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre-Luc Mouchel et al.
Cancers, 12(10) (2020-10-16)
Dendrogenin A (DDA), a mammalian cholesterol metabolite with tumor suppressor properties, has recently been shown to exhibit strong anti-leukemic activity in acute myeloid leukemia (AML) cells by triggering lethal autophagy. Here, we demonstrated that DDA synergistically enhanced the toxicity of
Harilaos Filippakis et al.
Scientific reports, 8(1), 14161-14161 (2018-09-23)
Tuberous Sclerosis Complex (TSC), a rare genetic disorder with mechanistic target of rapamycin complex 1 (mTORC1) hyperactivation, is characterized by multi-organ hamartomatous benign tumors including brain, skin, kidney, and lung (Lymphangioleiomyomatosis). mTORC1 hyperactivation drives metabolic reprogramming including glucose and glutamine
María Cecilia Gimenez et al.
Journal of virology, 95(6) (2020-12-29)
Infectious bursal disease virus (IBDV) is the archetypal member of the family Birnaviridae and the etiological agent of Gumboro disease, a highly contagious immunosuppressive infection of concern to the global poultry sector for its adverse health effects in chicks. Unlike
Jian-Kang Chen et al.
The Journal of clinical investigation, 125(6), 2429-2444 (2015-05-20)
Kidney size adaptively increases as mammals grow and in response to the loss of 1 kidney. It is not clear how kidneys size themselves or if the processes that adapt kidney mass to lean body mass also mediate renal hypertrophy
Muhammad Zaeem Noman et al.
Science advances, 6(18), eaax7881-eaax7881 (2020-06-05)
One of the major challenges limiting the efficacy of anti-PD-1/PD-L1 therapy in nonresponding patients is the failure of T cells to penetrate the tumor microenvironment. We showed that genetic or pharmacological inhibition of Vps34 kinase activity using SB02024 or SAR405

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service