Skip to Content
Merck
All Photos(1)

Key Documents

EHU034321

Sigma-Aldrich

MISSION® esiRNA

targeting human CP

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GAATGGACTGTCCCCAAAGAAGTAGGACCCACTAATGCAGATCCTGTGTGTCTAGCTAAGATGTATTATTCTGCTGTGGAACCCACTAAAGATATATTCACTGGGCTTATTGGGCCAATGAAAATATGCAAGAAAGGAAGTTTACATGCAAATGGGAGACAGAAAGATGTAGACAAGGAATTCTATTTGTTTCCTACAGTATTTGATGAGAATGAGAGTTTACTCCTGGAAGATAATATTAGAATGTTTACAACTGCACCTGATCAGGTGGATAAGGAAGATGAAGACTTTCAGGAATCTAATAAAATGCACTCCATGAATGGATTCATGTATGGGAATCAGCCGGGTCTCACTATGTGCAAAGGAGATTCGGTCGTGTGGTACTTATTCAGCGCCGGAAATGAGGCCGATGTACATGGAATATACTTTTCAGGAAACACATATCTGTGGAGAGGAGAACGGAGAGACACAGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... CP(1356) , CP(1356)

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei-Wen Wang et al.
International journal of molecular sciences, 20(18) (2019-09-25)
Wilson's disease (WD) is an autosomal recessive disorder of copper metabolism caused by defects in the ATPase gene (ATP7B). The various clinical features result from the massive accumulation of copper in the liver, cornea and basal ganglia. Although WD can
Yi Zhang et al.
Experimental cell research, 358(2), 234-241 (2017-07-01)
Neddylation inhibitor Pevonedistat (MLN4924) is a novel anticancer drug and has demonstrated broad-spectrum anticancer activity. Nevertheless, we found that Pevonedistat had only a modest apoptotic effect in osteosarcoma (OS) cells. Moreover, we noted that inhibition of neddylation by Pevonedistat led
Seong Hye Park et al.
Oncogene, 39(1), 136-150 (2019-08-30)
Hypoxia, or the deficiency of oxygen, in solid tumors is majorly responsible for the progression of cancer and remains unaffected by chemotherapy, but still requires definitive definition of the hypoxia signaling. Hypoxia disrupts the complete folding of mitochondrial proteins, leading
H-L Huang et al.
Oncogenesis, 6(7), e359-e359 (2017-07-12)
MUC1-C overexpression has been associated with the progression of pancreatic tumors by promoting the aggressive and metastatic phenotypes. As MUC1 is a STAT3 target gene, STAT3 plays a major role in regulating MUC1-C expression. In this study, we report an
Gang Liu et al.
Cell death & disease, 10(10), 688-688 (2019-09-20)
CELF6, a member of the CELF family of RNA-binding proteins, regulates muscle-specific alternative splicing and contributes to the pathogenesis of myotonic dystrophy (DM), however the role of CELF6 in cancer cell proliferation is less appreciated. Here, we show that the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service