Skip to Content
Merck
All Photos(1)

Key Documents

EHU033391

Sigma-Aldrich

MISSION® esiRNA

targeting human MGMT

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGGCTGAATGCCTATTTCCACCAGCCCGAGGCTATCGAAGAGTTCCCCGTGCCGGCTCTTCACCATCCCGTTTTCCAGCAAGAGTCGTTCACCAGACAGGTGTTATGGAAGCTGCTGAAGGTTGTGAAATTCGGAGAAGTGATTTCTTACCAGCAATTAGCAGCCCTGGCAGGCAACCCCAAAGCCGCGCGAGCAGTGGGAGGAGCAATGAGAGGCAATCCTGTCCCCATCCTCATCCCGTGCCACAGAGTGGTCTGCAGCAGCGGAGCCGTGGGCAACTACTCCGGAGGACTGGCCGTGAAGGAATGGCTTCTGGCCCATGAAGGCCACCGGTTGGGGAAGCCAGGCTTGGGAGGGAGCTCAGGTCTGGCAGGGGCCTGGCTCAAGGGAGCGGGAGCTACCTCGGGCTCCCCGCCTGCTGGCCGAAACTGAGTATGTGCAGTAGGATGGATGTTTGAGCGACACACACGTGTA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thatchawan Thanasupawat et al.
Molecular oncology, 11(8), 1078-1098 (2017-05-14)
The multikinase inhibitor and FDA-approved drug dovitinib (Dov) crosses the blood-brain barrier and was recently used as single drug application in clinical trials for GB patients with recurrent disease. The Dov-mediated molecular mechanisms in GB cells are unknown. We used
Xinwei Li et al.
Pathology oncology research : POR, 26(2), 707-714 (2019-02-04)
Primary central nervous system lymphoma (PCNSL) is an aggressive and rare subtype of non-Hodgkin lymphoma, arising exclusively in the CNS with a poor prognosis. Previous evidence has proved that MGMT was a promising target involving in TMZ resistance of PCNSL.
Jin Cheng et al.
Toxicology, 389, 67-73 (2017-07-20)
N-methyl-2,2-di(chloroethyl)amine (HN2) is a kind of bifunctional alkyltating agent, which can react with nucleophilic groups in DNA and/or protein to form HN2-bridged crosslinking of target molecules, such as DNA-protein crosslinkings (DPC). O
Lucio Tentori et al.
BMC cancer, 14, 151-151 (2014-03-07)
Chemoresistance of glioblastoma multiforme (GBM) has been attributed to the presence within the tumor of cancer stem cells (GSCs). The standard therapy for GBM consists of surgery followed by radiotherapy and the chemotherapeutic agent temozolomide (TMZ). However, TMZ efficacy is
Boswellic acid has anti-inflammatory effects and enhances the anticancer activities of Temozolomide and Afatinib, an irreversible ErbB family blocker, in human glioblastoma cells.
Manlio Barbarisi et al.
Phytotherapy research : PTR, 33(6), 1670-1682 (2019-03-30)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service