Skip to Content
Merck
All Photos(1)

Key Documents

EHU023381

Sigma-Aldrich

MISSION® esiRNA

targeting human FFAR2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCTGGTGCTCTTCTTCATCCCCATGGCAGTCACCATCTTCTGCTACTGGCGTTTTGTGTGGATCATGCTCTCCCAGCCCCTTGTGGGGGCCCAGAGGCGGCGCCGAGCCGTGGGGCTGGCTGTGGTGACGCTGCTCAATTTCCTGGTGTGCTTCGGACCTTACAACGTGTCCCACCTGGTGGGGTATCACCAGAGAAAAAGCCCCTGGTGGCGGTCAATAGCCGTGGTGTTCAGTTCACTCAACGCCAGTCTGGACCCCCTGCTCTTCTATTTCTCTTCTTCAGTGGTGCGCAGGGCATTTGGGAGAGGGCTGCAGGTGCTGCGGAATCAGGGCTCCTCCCTGTTGGGACGCAGAGGCAAAGACACAGCAGAGGGGACAAATGAGGACAGGGGTGTGGGTCAAGGAGAAGGGATGCCAAGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laure B Bindels et al.
British journal of cancer, 117(9), 1336-1340 (2017-09-06)
Activation of free fatty acid receptor 2 (FFAR2) by microbiota-derived metabolites (e.g., propionate) reduces leukaemic cell proliferation in vitro. This study aims to test whether Ffar2 expression per se also influences leukaemia cell growth in vivo. Bcr-Abl-expressing BaF cells were
Rebecca Roy et al.
Journal of molecular endocrinology, 65(2), 21-34 (2020-06-25)
Gestational diabetes mellitus (GDM) affects up to 16% of pregnant women and is associated with significant long-term health detriments for the mother and her offspring. Two central features of GDM are low-grade inflammation and maternal peripheral insulin resistance, therefore therapeutics
Hiroki Yoshida et al.
Archives of biochemistry and biophysics, 672, 108057-108057 (2019-07-30)
Short-chain fatty acids (SCFAs) such as acetate, propionate, and butyrate are generated by gut microbial fermentation of dietary fiber. SCFAs may exert multiple beneficial effects on human lipid and glucose metabolism. However, their actions and underlying mechanisms are not fully
Guangwen Wang et al.
Journal of virology, 94(2) (2019-11-07)
Influenza A virus (IAV) coopts numerous host factors to complete its replication cycle. Here, we identify free fatty acid receptor 2 (FFAR2) as a cofactor for IAV entry into host cells. We found that downregulation of FFAR2 or Ffar2 expression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service