Skip to Content
Merck
All Photos(1)

Key Documents

EHU020491

Sigma-Aldrich

MISSION® esiRNA

targeting human CFTR

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAGAATGGCCAACTCTCGAAAGTTATGATTATTGAGAATTCACACGTGAAGAAAGATGACATCTGGCCCTCAGGGGGCCAAATGACTGTCAAAGATCTCACAGCAAAATACACAGAAGGTGGAAATGCCATATTAGAGAACATTTCCTTCTCAATAAGTCCTGGCCAGAGGGTGGGCCTCTTGGGAAGAACTGGATCAGGGAAGAGTACTTTGTTATCAGCTTTTTTGAGACTACTGAACACTGAAGGAGAAATCCAGATCGATGGTGTGTCTTGGGATTCAATAACTTTGCAACAGTGGAGGAAAGCCTTTGGAGTGATACCACAGAAAGTATTTATTTTTTCTGGAACATTTAGAAAAAACTTGGATCCCTATGAACAGTGGAGTGATCAAGAAATATGGAAAGTTGCAGATGAGGTTGGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU131761EHU122191EHU029121
product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

AAGAATGGCCAACTCTCGAAAGTTATGATTATTGAGAATTCACACGTGAAGAAAGATGACATCTGGCCCTCAGGGGGCCAAATGACTGTCAAAGATCTCACAGCAAAATACACAGAAGGTGGAAATGCCATATTAGAGAACATTTCCTTCTCAATAAGTCCTGGCCAGAGGGTGGGCCTCTTGGGAAGAACTGGATCAGGGAAGAGTACTTTGTTATCAGCTTTTTTGAGACTACTGAACACTGAAGGAGAAATCCAGATCGATGGTGTGTCTTGGGATTCAATAACTTTGCAACAGTGGAGGAAAGCCTTTGGAGTGATACCACAGAAAGTATTTATTTTTTCTGGAACATTTAGAAAAAACTTGGATCCCTATGAACAGTGGAGTGATCAAGAAATATGGAAAGTTGCAGATGAGGTTGGG

esiRNA cDNA target sequence

CTGCTTTTCAAGCTGCTCCTGATCGGGGATTCCGGAGTGGGGAAGACCTGCGTCCTTTTTCGTTTTTCGGATGATGCCTTCAATACTACCTTTATTTCCACCATAGGAATAGACTTCAAGATCAAAACAGTTGAATTACAAGGAAAGAAGATCAAGCTACAGATATGGGATACAGCAGGCCAGGAGCGATTTCACACCATCACAACCTCCTACTACAGAGGCGCAATGGGTATCATGCTAGTATATGACATCACCAATGGTAAAAGTTTTGAAAACATCAGCAAATGGCTTAGAAACATAGATGAGCATGCCAATGAAGATGTGGAAAGAATGTTACTAGGAAACAAGTGTGATATGGACGACAAAAGAGTTGTACCTAAAGGAAAAGGAGAACAGATTGCAAGGGA

esiRNA cDNA target sequence

CCCACAGTTCCTCTGGACTTCTCTGGACCACAGTCCTCTGCCAGACCCCTGCCAGACCCCAGTCCACCATGATCCATCTGGGTCACATCCTCTTCCTGCTTTTGCTCCCAGTGGCTGCAGCTCAGACGACTCCAGGAGAGAGATCATCACTCCCTGCCTTTTACCCTGGCACTTCAGGCTCTTGTTCCGGATGTGGGTCCCTCTCTCTGCCGCTCCTGGCAGGCCTCGTGGCTGCTGATGCGGTGGCATCGCTGCTCATCGTGGGGGCGGTGTTCCTGTGCGCACGCCCACGCCGCAGCCCCGCCCAAGAAGATGGCAAAGTCTACATCAACATGCCAGGCAGGGGCTGACCCTCCTGCAGCTTGGACCTTTGACTTCTGACCCTCTCATCCTGG

esiRNA cDNA target sequence

ACTCCTTTGAGCTGCTGTCCCACACCGTGGAAGAGAAGATTATCATTGACAAGGAATATTATTACACCAAGGAGGAGCAGTTTAAACAGGTGTTCAAGGAGCTGGAGTTTCTGGGTTGGTATACCACAGGGGGGCCACCTGACCCCTCGGACATCCACGTCCATAAGCAGGTGTGTGAGATCATCGAGAGCCCCCTCTTTCTGAAGTTGAACCCTATGACCAAGCACACAGATCTTCCTGTCAGCGTTTTTGAGTCTGTCATTGATATAATCAATGGAGAGGCCACAATGCTGTTTGCTGAGCTGACCTACACTCTGGCCACAGAGGAAGCGGAACGCATTGGTGTAGACCACGTAGCCCGAATGACAGCAACAGGCAGTGGAGAGAACTCCACTGTGGCTGAACACCTGATAGCACAGCACAGCGCCATCAAGATGCTGCACAGCCGCGTCAAGCTCATCTTGGAGTACGTCAAGGCCTCTG

Ensembl | human accession no.

ENSG00000001626

Ensembl | human accession no.

ENSG00000084733

Ensembl | human accession no.

ENSG00000126264

Ensembl | human accession no.

ENSG00000168090

NCBI accession no.

NM_000492

NCBI accession no.

NM_016131

NCBI accession no.

NM_001007469

NCBI accession no.

NM_006833

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luiz Felipe M Prota et al.
PloS one, 7(12), e47405-e47405 (2012-12-29)
Dexamethasone is widely used for pulmonary exacerbation in patients with cystic fibrosis, however, not much is known about the effects of glucocorticoids on the wild-type cystic fibrosis channel transmembrane regulator (CFTR). Our aim was to determine the effects of dexamethasone
Babita Kaundal et al.
Journal of materials chemistry. B, 8(37), 8658-8670 (2020-08-28)
Acute myeloid leukemia (AML), which is common in the elderly population, accounts for poor long-term survival with a high possibility of relapse. The associated lack of currently developed therapeutics is directing the search for new therapeutic targets relating to AML.
Maria Favia et al.
Molecular biology of the cell, 21(1), 73-86 (2009-11-06)
We have demonstrated that Na(+)/H(+) exchanger regulatory factor 1 (NHERF1) overexpression in CFBE41o- cells induces a significant redistribution of F508del cystic fibrosis transmembrane conductance regulator (CFTR) from the cytoplasm to the apical membrane and rescues CFTR-dependent chloride secretion. Here, we
C Norez et al.
British journal of pharmacology, 171(21), 4831-4849 (2014-07-30)
The most common mutation in cystic fibrosis (CF), F508del, causes defects in trafficking, channel gating and endocytosis of the CF transmembrane conductance regulator (CFTR) protein. Because CF is an orphan disease, therapeutic strategies aimed at improving mutant CFTR functions are
Yonghwan Shin et al.
Journal of clinical medicine, 9(3) (2020-03-19)
Cystic fibrosis transmembrane conductance regulator (CFTR), a cyclic AMP (cAMP)-regulated chloride channel, is critical for secretion and absorption across diverse epithelia. Mutations or absence of CFTR result in pathogeneses, including cancer. While CFTR has been proposed as a tumor suppressing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service