Skip to Content
Merck

EHU014361

Sigma-Aldrich

MISSION® esiRNA

targeting human EBP

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Availability
Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Availability
Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTGCAGTGTGTGGGTTCATTCACCTGGTGATCGAGGGCTGGTTCGTTCTCTACTACGAAGACCTGCTTGGAGACCAAGCCTTCTTATCTCAACTCTGGAAAGAGTATGCCAAGGGAGACAGCCGATACATCCTGGGTGACAACTTCACAGTGTGCATGGAAACCATCACAGCTTGCCTGTGGGGACCACTCAGCCTGTGGGTGGTGATCGCCTTTCTCCGCCAGCATCCCCTCCGCTTCATTCTACAGCTTGTGGTCTCTGTGGGCCAGATCTATGGGGATGTGCTCTACTTCCTGACAGAGCACCGCGACGGATTCCAGCACGGAGAGCTGGGCCACCCTCTCTACTTCTGGTTTTACTTTGTCTTCATGAATGCCCTGTGGCTGGTGCTGCCTGGAGTCCTTGTGCTTGATGCTGTGAAGCACCTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU016631EHU002631EHU010781
esiRNA cDNA target sequence

TTGCAGTGTGTGGGTTCATTCACCTGGTGATCGAGGGCTGGTTCGTTCTCTACTACGAAGACCTGCTTGGAGACCAAGCCTTCTTATCTCAACTCTGGAAAGAGTATGCCAAGGGAGACAGCCGATACATCCTGGGTGACAACTTCACAGTGTGCATGGAAACCATCACAGCTTGCCTGTGGGGACCACTCAGCCTGTGGGTGGTGATCGCCTTTCTCCGCCAGCATCCCCTCCGCTTCATTCTACAGCTTGTGGTCTCTGTGGGCCAGATCTATGGGGATGTGCTCTACTTCCTGACAGAGCACCGCGACGGATTCCAGCACGGAGAGCTGGGCCACCCTCTCTACTTCTGGTTTTACTTTGTCTTCATGAATGCCCTGTGGCTGGTGCTGCCTGGAGTCCTTGTGCTTGATGCTGTGAAGCACCTC

esiRNA cDNA target sequence

GCCACACTTACCTTGCCAATGGCCAGACCAAGGTGCTGACTCAGAAGTTGTCATCAGTCAGAGGCAATCATATTATCTCAGGGACATGCGCATCATGGCGTGGCAAGAGCCTTCGGGTTCAGAAGATCAGGCCTTCTATCCTGGGCTGCAACATCCTTCGAGTTGAATATTCCTTACTGATCTATGTTAGCGTTCCTGGATCCAAGAAGGTCATCCTTGACCTGCCCCTGGTAATTGGCAGCAGATCAGGTCTAAGCAGCAGAACATCCAGCATGGCCAGCCGAACCAGCTCTGAGATGAGTTGGGTAGATCTGAACATCCCTGATACCCCAGAAGCTCCTCCCTGCTATATGGATGTCATTCCTGAAGATCACCGATTGGAGAGCCCAACCACTCCTCTGCTAGATGACATGGATGGCTCTCAAG

esiRNA cDNA target sequence

CCAGGACCTCTTCACAGAGCTGAAAAGGTACTACACTGGGGGTAATGTGAATCTGGAGGAAATGCTCAATGACTTTTGGGCTCGGCTCCTGGAACGGATGTTTCAGCTGATAAACCCTCAGTATCACTTCAGTGAAGACTACCTGGAATGTGTGAGCAAATACACTGACCAGCTCAAGCCATTTGGAGACGTGCCCCGGAAACTGAAGATTCAGGTTACCCGCGCCTTCATTGCTGCCAGGACCTTTGTCCAGGGGCTGACTGTGGGCAGAGAAGTTGCAAACCGAGTTTCCAAGGTCAGCCCAACCCCAGGGTGTATCCGTGCCCTCATGAAGATGCTGTACTGCCCATACTGTCGGGGGCTTCCCACTGTGAGGCCCTGCAACAACTACTGTCTCAACGTCATGAAGGGCTGCTTGGCAAATCAGGCTGACCTCGACAC

esiRNA cDNA target sequence

CCTGAGCATGATTCAGAGCAAGTGCAGTGTGCCCTTCACCCCTATTGAGTTTCACTATGAGAATACACGGGCCCAGTTCTTCGTTGAAGACGCCAGTACTGCCTCTGCATTGAAGGCTGTCAACTATAAGATTTTGGATCGGGAGAACCGAAGGATATCTATCATCATCAACTCTTCTGCTCCACCCCACACTATACTGAATGAACTGAAGCCAGAACAAGTAGAACAGCTAAAGCTGATCATGAGCAAACGATACGATGGCTCCCAACAAGCCCTTGACCTCAAAGGCCTCCGTTCAGACCCAGATTTGGTGGCCCAGAACATTGACGTTGTCCTGAATCGCAGAAGCTGTATGGCAGCTACCCTGAGGATCATTGAAGAGAACATCCCTGAGCTATTGTCCTTGAACTTGAGCAACAACAGGCTGTACAGGCTGGATGACATGTCTAGCATTGTTCAGAAGGCACCCAACCTGAAGATCCTAAACCTTTCTGGAAATGAATTGAAGTCTGAGCGGGAATT

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Gene Information

human ... EBP(10682), EBP(10682)

Gene Information

human ... TXNIP(10628), TXNIP(10628)

Gene Information

human ... GPC6(10082), GPC6(10082)

Gene Information

human ... NXF1(10482), NXF1(10482)

NCBI accession no.

NM_006579

NCBI accession no.

NM_006472

NCBI accession no.

NM_005708

NCBI accession no.

NM_006362

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service