Skip to Content
Merck
All Photos(1)

Key Documents

EHU008741

Sigma-Aldrich

MISSION® esiRNA

targeting human PLXNA1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGTGGAGAAGTCGCTGACACTGTTCGGGCAGCTGCTGACCAAGAAGCACTTCCTGCTGACCTTCATCCGCACGCTGGAGGCACAGCGCAGCTTCTCCATGCGCGACCGCGGGAATGTGGCCTCGCTCATCATGACGGCCCTGCAGGGCGAGATGGAATACGCCACAGGCGTGCTCAAGCAGCTGCTTTCCGACCTCATCGAGAAGAACCTGGAGAGCAAGAACCACCCCAAGCTGCTACTGCGCCGGACTGAGTCGGTGGCAGAGAAGATGCTAACTAACTGGTTCACCTTCCTCTTGTATAAGTTCCTCAAGGAGTGCGCTGGGGAGCCGCTGTTCATGCTGTACTGCGCCATCAAGCAGCAGATGGAGAAGGGCCCCATTGACGCCATCACGGGTGAGGCACGCTACTCCCTGAGTGAGGACAAGCTCATCCG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Wei Wang et al.
EBioMedicine, 46, 66-78 (2019-08-07)
MicroRNAs (miRNAs) are involved in oncogenesis of esophageal squamous cell carcinoma (ESCC). miR-134 is reported to have a tumour-suppressive role but its role in ESCC is not known. The present study was designed to examine whether miR-134 inhibits ESCC development
Jason R Dobson et al.
Cancer cell international, 14, 73-73 (2014-08-15)
For treatment and prevention of metastatic disease, one of the premier challenges is the identification of pathways and proteins to target for clinical intervention. Micro RNAs (miRNAs) are short, non-coding RNAs, which regulate cellular activities by either mRNA degradation or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service