Skip to Content
Merck
All Photos(1)

Key Documents

EHU007001

Sigma-Aldrich

MISSION® esiRNA

targeting human DSP

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGAAGTCGTTGTTGGCCACTATGAAGACAGAACTACAGAAAGCCCAGCAGATCCACTCTCAGACTTCACAGCAGTATCCACTTTATGATCTGGACTTGGGCAAGTTCGGTGAAAAAGTCACACAGCTGACAGACCGCTGGCAAAGGATAGATAAACAGATCGACTTTAGGTTATGGGACCTGGAGAAACAAATCAAGCAATTGAGGAATTATCGTGATAACTATCAGGCTTTCTGCAAGTGGCTCTATGATGCTAAACGCCGCCAGGATTCCTTAGAATCCATGAAATTTGGAGATTCCAACACAGTCATGCGGTTTTTGAATGAGCAGAAGAACTTGCACAGTGAAATATCTGGCAAACGAGACAAATCAGAGGAAGTACAAAAAATTGCTGAACTTTGCGCCAATTCAATTAAGGATTATGAGCTCCAGCTGGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peipei Li et al.
International journal of biological sciences, 15(11), 2350-2362 (2019-10-09)
The interaction between genomic DNA and protein fundamentally determines the activity and the function of DNA elements. Capturing the protein complex and identifying the proteins associated with a specific DNA locus is difficult. Herein, we employed CRISPR, the well-known gene-targeting
Aritro Nath et al.
Molecular cancer research : MCR, 19(2), 240-248 (2020-10-28)
Elevated uptake of saturated fatty acid palmitate is associated with metastatic progression of cancer cells; however, the precise signaling mechanism behind the phenomenon is unclear. The loss of cell adhesion proteins, such as desmoplakin (DSP), is a key driving event
Minnah Thomas et al.
Current biology : CB, 30(4), 682-690 (2020-02-01)
During apoptosis, or programmed cell death, a dead cell could be expelled from the tissue by coordinated processes between the dying cell and its neighbors. Apoptotic cell extrusion is driven by actomyosin cable formation and its contraction and lamellipodial crawling
Dipal M Patel et al.
The Journal of cell biology, 206(6), 779-797 (2014-09-17)
Mechanisms by which microtubule plus ends interact with regions of cell-cell contact during tissue development and morphogenesis are not fully understood. We characterize a previously unreported interaction between the microtubule binding protein end-binding 1 (EB1) and the desmosomal protein desmoplakin
Johan Sternemalm et al.
PloS one, 10(8), e0134789-e0134789 (2015-08-05)
Deleterious mutations of the Centrosome/Spindle Pole associated Protein 1 gene, CSPP1, are causative for Joubert-syndrome and Joubert-related developmental disorders. These disorders are defined by a characteristic mal-development of the brain, but frequently involve renal and hepatic cyst formation. CSPP-L, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service