Skip to Content
Merck
All Photos(1)

Key Documents

EHU005981

Sigma-Aldrich

MISSION® esiRNA

targeting human EPS8

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGAATGGCTACGGATCATCACCTACCTTTTCCCAGACGGACAGAGAACATGGTTCAAAAACAAGTGCAAAGGCCCTTTATGAACAAAGGAAGAATTATGCACGGGACAGTGTCAGCAGTGTGTCAGATATATCTCAATACCGTGTTGAACACTTGACTACCTTTGTCCTGGATCGGAAAGATGCTATGATCACTGTTGATGATGGAATAAGGAAATTGAAATTGCTTGATGCCAAGGGCAAAGTGTGGACTCAAGATATGATTCTTCAAGTGGATGACAGAGCTGTGAGCCTGATTGATTTAGAATCAAAGGCAAGTAATGAACTGGAGAATTTTCCTTTAAACACAATCCAGCACTGCCAAGCTGTGATGCATTCATGCAGCTATGATTCAGTTCTTGCACTGGTGTGCAAAGAGCCAACCCAGAACAAGCCAGAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jieyun Zhang et al.
Acta biochimica et biophysica Sinica, 52(3), 259-267 (2020-03-10)
Tumor metastasis is the main cause of treatment failure and death in patients with late stage of gastric cancer (GC). Studies showed that microRNAs (miRNAs) are important regulators in the process of tumor metastasis. In this study, we used miRNA
Huifang Lu et al.
Molecular medicine reports, 14(6), 4999-5006 (2016-11-15)
Epidermal growth factor receptor pathway substrate 8 (EPS8) is critical in the proliferation, progression and metastasis of solid and hematological types of cancer, and thus constitutes an ideal target for cancer immunotherapy. The present study aimed to identify human leukocyte antigen
Quan Yuan et al.
International journal of pharmaceutics, 557, 178-181 (2019-01-01)
We developed polyamidoamine dendrimers conjugated with epidermal growth factor (EGF) for use in receptor-mediated delivery of therapeutics to cancer cells. Here, we demonstrate the utility of this approach to inhibit proliferation and migration of head and neck squamous carcinoma cells
Haruhi Fukuhisa et al.
Journal of human genetics, 64(6), 521-534 (2019-03-13)
Our ongoing analyses identifying dysregulated microRNAs (miRNAs) and their controlled target RNAs have shed light on novel oncogenic pathways in pancreatic ductal adenocarcinoma (PDAC). The PDAC miRNA signature obtained by RNA sequencing showed that both strands of pre-miR-130b (miR-130b-5p, the
Elisa Cappellini et al.
Life sciences, 131, 30-36 (2015-04-22)
Eps8 is an actin-binding protein which has been proposed as a regulator of cancer cell motility and invasion. However, nothing much is known about its contribution to the invasive properties of endothelial cells (ECs), and more generally to angiogenesis. Expression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service