Skip to Content
Merck
All Photos(1)

Documents

EHU004571

Sigma-Aldrich

MISSION® esiRNA

targeting human ASPN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATTCCTGGCTTTGTGCTCTGCCAAACCCTTCTTTAGCCCTTCACACATCGCACTGAAGAATATGATGCTGAAGGATATGGAAGACACAGATGATGATGATGATGATGATGATGATGATGATGATGATGATGAGGACAACTCTCTTTTTCCAACAAGAGAGCCAAGAAGCCATTTTTTTCCATTTGATCTGTTTCCAATGTGTCCATTTGGATGTCAGTGCTATTCACGAGTTGTACATTGCTCAGATTTAGGTTTGACCTCAGTCCCAACCAACATTCCATTTGATACTCGAATGCTTGATCTTCAAAACAATAAAATTAAGGAAATCAAAGAAAATGATTTTAAAGGACTCACTTCACTTTATGGTCTGATCCTGAACAACAACAAGCTAACGAAGATTCACCCAAAAGCCTTTCTAACCACAAAGAAGTTGCGAAGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Li Li et al.
Cell biology international (2019-09-10)
The progression of diabetic cardiomyopathy is related to cardiomyocyte dysfunction and apoptosis. Our previous studies showed that asporin (ASPN) was significantly increased in the myocardium of db/db mice through proteomics, and grape seed procyanidin B2 (GSPB2) significantly inhibited the expression
Hengcun Li et al.
Cell death & disease, 10(2), 109-109 (2019-02-08)
Previous studies revealed that Asporin (ASPN) is a potential mediator in the development of various types of cancer as a secreted stroma protein, but the function of ASPN inside the cancer cells remains largely unknown. Here, we demonstrated a higher

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service