Skip to Content
Merck
All Photos(1)

Documents

EHU003861

Sigma-Aldrich

MISSION® esiRNA

targeting human CDKN1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACACCACTGGAGGGTGACTTCGCCTGGGAGCGTGTGCGGGGCCTTGGCCTGCCCAAGCTCTACCTTCCCACGGGGCCCCGGCGAGGCCGGGATGAGTTGGGAGGAGGCAGGCGGCCTGGCACCTCACCTGCTCTGCTGCAGGGGACAGCAGAGGAAGACCATGTGGACCTGTCACTGTCTTGTACCCTTGTGCCTCGCTCAGGGGAGCAGGCTGAAGGGTCCCCAGGTGGACCTGGAGACTCTCAGGGTCGAAAACGGCGGCAGACCAGCATGACAGATTTCTACCACTCCAAACGCCGGCTGATCTTCTCCAAGAGGAAGCCCTAATCCGCCCACAGGAAGCCTGCAGTCCTGGAAGCGCGAGGGCCTCAAAGGCCCGCTCTACATCTTCTGCCTTAGTCTCAGTTTGTGTGTCTTAATTATTATTTGTGTTTTAATTTAAACACCTCCTCATGTACATACCCTGGCCGCCCCCTGCCCCCCAGCCTCTGGCATTAGAATTATTTAAACAAAAACTAGGCGGTTGAATGAGAGGTTCCTAAGAGTGCTGGGCAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Ji et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 40(5), 895-907 (2016-12-13)
The Notch signaling pathway has been implicated in the pericyte phenotype, but its exact roles in hemangioma-derived pericytes (Hem-pericytes) remain ill defined. Hem-pericytes were stimulated by immobilized recombinant Jagged1. The potential mechanisms of Notch-induced Hem-pericytes growth arrest were investigated by
Anna E Vilgelm et al.
EBioMedicine, 24, 43-55 (2017-10-17)
Antagonists of MDM2-p53 interaction are emerging anti-cancer drugs utilized in clinical trials for malignancies that rarely mutate p53, including melanoma. We discovered that MDM2-p53 antagonists protect DNA from drug-induced damage in melanoma cells and patient-derived xenografts. Among the tested DNA
Mugdha Patki et al.
Scientific reports, 8(1), 16006-16006 (2018-10-31)
Dexamethasone (Dex), co-administered to lung adenocarcinoma patients with pemetrexed chemotherapy, protects against pemetrexed cytotoxicity by inducing reversible G1 arrest, reflected by the effect of Dex on FLT-PET images of patient tumors. However, perioperative Dex treatment increases survival but the mechanism
Hui Zhu et al.
Stem cells (Dayton, Ohio), 32(8), 2098-2110 (2014-04-18)
In mammalian embryos, embryonic stem cells (ESCs) and induced pluripotent cells, a shortened G1 phase is correlated with the pluripotent state. To molecularly define this phase, we compared transcripts from the shortened G1 of human ESCs (hESCs) with those from
Outhiriaradjou Benard et al.
Molecular cancer research : MCR, 17(7), 1571-1581 (2019-04-11)
Cancer stem cells (CSC) generate and sustain tumors due to tumor-initiating potential, resulting in recurrence or metastasis. We showed that knockout of the cell-cycle inhibitor, p21CIP1, in the PyMT mammary tumor model inhibits metastasis; however the mechanism remained unknown. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service