Skip to Content
Merck
All Photos(1)

Key Documents

EHU002001

Sigma-Aldrich

MISSION® esiRNA

targeting human HDAC2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGTTGCTGGAGCTGTGAAGTTAAACCGACAACAGACTGATATGGCTGTTAATTGGGCTGGAGGATTACATCATGCTAAGAAATCAGAAGCATCAGGATTCTGTTACGTTAATGATATTGTGCTTGCCATCCTTGAATTACTAAAGTATCATCAGAGAGTCTTATATATTGATATAGATATTCATCATGGTGATGGTGTTGAAGAAGCTTTTTATACAACAGATCGTGTAATGACGGTATCATTCCATAAATATGGGGAATACTTTCCTGGCACAGGAGACTTGAGGGATATTGGTGCTGGAAAAGGCAAATACTATGCTGTCAATTTTCCAATGAGAGATGGTATAGATGATGAGTCATATGGGCAGATATTTAAGCCTATTATCTCAAAGGTGATGGAGATGTATCAACCTAGTGCTGTGGTATTACAGTGTGGTGCAGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David B Wang et al.
Brain pathology (Zurich, Switzerland), 29(2), 164-175 (2018-07-22)
Histone deacetylases (HDACs) catalyze acetyl group removal from histone proteins, leading to altered chromatin structure and gene expression. HDAC2 is highly expressed in adult brain, and HDAC2 levels are elevated in Alzheimer's disease (AD) brain. We previously reported that neuron-specific
Jie Gao et al.
Gene, 678, 1-7 (2018-08-01)
Chronic diabetic foot ulcer (DFU) is a major cause of disability and mortality in patients with diabetes. Dysfunctional endothelial progenitor cells (EPCs) play important roles in preventing vascular complications in these patients. Our results determined the elevated expression of histone
Wen-Feng Fang et al.
Journal of inflammation (London, England), 15, 3-3 (2018-01-19)
Sepsis is a life-threatening organ dysfunction caused by dysregulated host response to infection, and is primarily characterized by an uncontrolled systemic inflammatory response. In the present study, we developed an effective adjunct therapy mediated by a novel mechanism, to attenuate
Ziran Wang et al.
Molecular therapy oncolytics, 17, 547-561 (2020-07-09)
Hepatocellular carcinoma (HCC) is a common malignant tumor. LukS-PV is the S component of Panton-Valetine leukocidin (PVL), which is secreted by Staphylococcus aureus. This study investigated the effects of LukS-PV on the proliferation, apoptosis, and cell-cycle progression of HCC cells
Sin Y Choi et al.
Journal of cellular and molecular medicine, 20(12), 2289-2298 (2016-07-16)
Epithelial-mesenchymal transition (EMT) and renal fibrosis are closely involved in chronic kidney disease. Inhibition of histone deacetylase (HDAC) has an anti-fibrotic effect in various diseases. However, the pathophysiological role of isoform-specific HDACs or class-selective HDACs in renal fibrosis remains unknown.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service