Skip to Content
Merck
All Photos(1)

Key Documents

EHU000091

Sigma-Aldrich

MISSION® esiRNA

targeting human DRD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCTCTGATGGAAATGCCACTTCCCTGGCTGAGACCATAGACAACTGTGACTCCAGCCTCAGCAGGACATATGCCATCTCATCCTCTGTAATAAGCTTTTACATCCCTGTGGCCATCATGATTGTCACCTACACCAGGATCTACAGGATTGCTCAGAAACAAATACGGCGCATTGCGGCCTTGGAGAGGGCAGCAGTCCACGCCAAGAATTGCCAGACCACCACAGGTAATGGAAAGCCTGTCGAATGTTCTCAACCGGAAAGTTCTTTTAAGATGTCCTTCAAAAGAGAAACTAAAGTCCTGAAGACTCTGTCGGTGATCATGGGTGTGTTTGTGTGCTGTTGGCTACCTTTCTTCATCTTGAACTGCATTTTGCCCTTCTGTGGGTCTGGGGAGACGCAGCCCTTCTGCATTGATTCCAACACCTTTGACGTGTTTGTGTGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengzhi Liu et al.
Cancer research, 78(14), 3865-3876 (2018-05-18)
While bone is a frequent target of breast cancer-associated metastasis, little is known about the effects of tumor-bone interactions on the efficacy of tumor-suppressing agents. Here we examined the effect of two FDA-approved dopamine modulators, fluphenazine and trifluoperazine, on mammary
Kazumasa Minami et al.
Scientific reports, 7, 45686-45686 (2017-04-05)
Dopaminergic signaling plays a critical role in the nervous system, but little is known about its potential role in breast cancer and bone metabolism. A screening of ~1,000 biologically active compounds revealed that a selective agonist of dopamine receptor D1
Fengmin Li et al.
Endocrinology, 156(6), 2211-2221 (2015-04-01)
Sorting nexin 5 (SNX5) belongs to the SNX family, which is composed of a diverse group of proteins that mediate trafficking of plasma membrane proteins, receptors, and transporters. SNX5 is important in the resensitization of the dopamine D1-like receptor (D1R).
Kang Yang et al.
Cellular oncology (Dordrecht), 43(6), 1175-1190 (2020-08-08)
Recent studies have reported important roles of dopamine receptors in the early development and progression of glioblastoma (GBM). Here, we tested the antitumor activity of a Dopamine receptor D1 (DRD1) agonist, either alone or in combination with temozolomide (TMZ) on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service