Skip to Content
Merck

Skip To

EC896

GenElute-E Viral RNA/DNA Swab 96 kit

Reagents and consumables for viral RNA and DNA purification. 96-well format.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41105600
NACRES:
NB.23
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

input

DNA
RNA

storage temp.

2-8°C

General description

GenElute-E purification kits provide a simple alternative for viral RNA and DNA purification. Compared with traditionalmethods, GenElute-E offers:
  • Isolation of viral RNA and DNA in less than 30 minutes with minimal sample handling
  • Better purity, leading to improved performance in PCR and other applications
  • Significant reduction in plastic and hazardous chemical waste

Traditional silica-based, bind-wash-elute purification kits require multiple wash steps to remove impurities from the spin columns. These steps increase the risk of cross contamination, and subject the nucleic acids to centrifugation sheering forces.

GenElute-E kits employ size exclusion negative chromatography to separate large nucleic acid molecules from smaller protein, lipid, and ionic components in swab, cell, tissue, blood, and other samples. Single-spin columns efficiently absorb and retain cellular debris and sample contaminantswhile allowing nucleic acids to pass through, reducing the number of steps and plastic materials required for purification. This novel method for high-quality purification is made possible by our innovative SmartLyse® Buffer, which enables fast and efficient isolation.

Isolate nucleic acid in a fraction of time compared to traditional silica bind-wash-elute procedures. Simply prepare spin column, mix sample with lysis components, pipette sample onto the spin column, and centrifuge directly into a collection tube. Cellular debris and contaminants remain bound in the column to be discarded, while purified viral RNA and DNA is ready to be used in downstream applications or stored.

Application

Suitable for most common downstream applications, including genotyping, PCR, and NGS
The GenElute-E Viral RNA/DNA Swab kit has been developed to purify genomic viral RNA and DNA from nasopharyngeal swabs, dry or in transport medium, in addition to genital swabs and stool samples.

The sample lysis utilizes the SmartLyse® Viral Buffer for fast and efficient isolation of nucleic acids from a variety of swab types and stool samples.

Purified viral RNA and DNA is eluted in Tris buffer, pH 7.8 and can immediately be used for most common downstream applications, including PCR, genotyping, NGS, and others. Final yield is comparable to most common silica-based methods. GenElute-E purified genomic DNA preparations commonly show an A260/280 ratio of around 1.8.

Features and Benefits

  • Fast and efficient purification: no lysis incubation and minimal hands-on, preparation steps
  • Higher quality nucleic acids: negative chromotography-based purification reduces carryover of PCR inhibitors and other contaminents
  • Reduced environmental impact: 55% less plastic waste, as well as a significant reduction in hazardous chemicals

Other Notes

We believe that green chemistry will contribute to a better tomorrow. With a growing portfolio of greener alternatives, there are now more choices to reduce the ecological impact of your research while still delivering quality and efficacy so your results are not compromised. The single-spin protocol limits the number of collection and pipetting steps, reducing plastic waste by 55% compared to equivalent bind-wash-elute methods. GeGenElute-E is designated a Greener Alternative Product based on the ""Prevention"" and ""Designing Safer Chemicals"" qualifications of The 12 Principles of Green Chemistry.GenElute-E kits also adhere to the principles of SMASH Packaging, our global initiative to improve the sustainability through the reduction of packaging materials, the increased use of sustainable alternatives, and improved recyclability. Packaging is FCS-marked for sustainable forestry practices and produced from greater than 70% recycled content.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC
SmartLyse is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EC848EC810EC996
storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp

input

DNA
RNA

input

DNA
RNA

input

sample(s) (swabs: nasopharyngeal and genital)
sample(s) (stool)

input

RNA
DNA

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Efecto del método de purificación en la precisión de la cuantificación del ADN y en los procesos enzimáticos posteriores Evaluación de la pureza del ADN genómico mediante espectrofotometría UV, electroforesis en gel y qPCR consecutiva utilizando los kits de purificación de ADN GenElute™-E.

GenElute™-E DNA purification kits ensure accurate DNA quantitation and downstream enzymatic processes.

Related Content

Animation showing technology principle of GenElute™-E single spin negative chromatography nucleic acid purification kits

Demo video showing how to purify DNA and RNA GenElute™-E single spin nucleic acid purification kits

Answers to frequently asked questions related to GenElute™-E single spin DNA and RNA purification and negative chromatography

View All Related Content




Questions

1–4 of 4 Questions  
  1. Do we know how stable the purified DNA is through several freeze-thaw cycles?

    1 answer
    1. This will fluctuate due to sample variability (sample collection, concentration, fragment length, sequence [GC content], storage before isolation, etc.). However, the sample is buffer exchanged into a standard storage buffer that is included in the kit (1X TE).

      Helpful?

  2. Genelute-e single spin DNA and RNA purification kits use negative chromatographty to isolate nucleic acids. Can you explain how this approach simplifies workflows?

    1 answer
    1. Instead of optimizing the bind, wash, and release steps in conventional silica-based spin purification preps, the technology focuses on a separation by performing a single step fractionation based on the size of the biomolecules, which results in depleted impurities.

      Helpful?

  3. What is the composition of the lysis buffer and clearing buffer after flowing through the resin?

    1 answer
    1. The presence of EDTA, SDS, or excess salt can affect my PCR/ sequencing reaction. The lysis buffer information is proprietary, but we can say it is free of chaotropic salts. The resins are desalting resins so EDTA, SDS, and salts are depleted.

      Helpful?

  4. Does the technology introduce any bias into the sample?

    1 answer
    1. GenElute™-E does not introduce biases that some “bind-wash-elute” technologies can add because the technology separates by size, rather than by what binds and what is released.

      Helpful?

Reviews

No rating value

Active Filters